Skip to Content
Merck
All Photos(1)

Key Documents

P8804

Sigma-Aldrich

Phospholipase C, Phosphatidylinositol-specific from Bacillus cereus

buffered aqueous glycerol solution, ≥1,000 units/mg protein (Lowry)

Synonym(s):

1-Phosphatidyl-D-myo-inositol inositol phosphohydrolase, cyclic-phosphate forming, PI-PLC

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

Bacillus sp. (Bacillus cereus)

Quality Level

form

buffered aqueous glycerol solution

specific activity

≥1,000 units/mg protein (Lowry)

mol wt

28 kDa

foreign activity

Phopholipase C (lecithinase) ≤1 units/mg protein
Sphingomyelinase ≤40 units/mg protein

storage temp.

2-8°C

Gene Information

Bacillus cereus E33L ... BCZK3513(3026815)

Looking for similar products? Visit Product Comparison Guide

General description

Phospholipase C (PI-PLC) from Bacillus cereus has irregular triosephosphate isomerase (TIM)-barrel structure with eight-standard parallel β barrel.[1] It is a 28 kDa protein.[2]

Application

Phospholipase C, Phosphatidylinositol-specific from Bacillus cereus has been used:
  • in the hydrolysis of substrates p-nitrophenylphosphorylcholine (p-NPPC) and p-nitrophenylphosphorylphosphate (p-NPP)[3]
  • to cleave glycosylphosphatidylinositol (GPI) anchor of lynx1 protein and its detachment from plasma membrane[4]
  • to cleave immunolabeled HeLa cells[5]

Biochem/physiol Actions

Phospholipase C (PI-PLC) releases diacylglycerol by cleaving the glycosylphosphatidylinositol.[1] It has broad substrate specificity and its activity is influenced by metal ions and surfactants.[2]
Used for the release of GPI anchored proteins from the membrane.

Unit Definition

One unit will liberate one unit of acetylcholinesterase per minute from a membrane-bound crude preparation at pH 7.4 at 30 °C (10 minute incubation).

Physical form

Solution in 60% (v/v) glycerol containing 10 mM Tris-HCl, pH 8.0 and 10 mM EDTA

Analysis Note

Acetylcholinesterase is measured according to Ellman, et al.

inhibitor

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ligand binding characteristics of a glycosylphosphatidyl inositol membrane-anchored HeLa cell folate receptor epitope-related to human milk folate binding protein
Holm J, et al.
Bioscience Reports, 20(2), 109-118 (2000)
High-level expression of recombinant phospholipase C from Bacillus cereus in Pichia pastoris and its characterization
Seo KH and Rhee JI
Biotechnology Letters, 26(19), 1475-1479 (2004)
Crystal structure of phosphatidylinositol-specific phospholipase C from Bacillus cereus in complex with glucosaminyl (alpha1? 6)-d-myo-inositol, an essential fragment of GPI anchors
Heinz DW, et al.
Biochemistry, 35(29), 9496-9504 (1996)
Lynx1 shifts alpha4beta2 nicotinic receptor subunit stoichiometry by affecting assembly in the endoplasmic reticulum
Nichols WA, et al.
The Journal of Biological Chemistry, 289(45), 31423-31432 (2014)
Critical evaluation of p-nitrophenylphosphorylcholine (p-NPPC) as artificial substrate for the detection of phospholipase C
Flieger A, et al.
Enzyme and Microbial Technology, 26(5-6), 451-458 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service