Skip to Content
Merck
All Photos(4)

Key Documents

P6621

Sigma-Aldrich

Phospholipase C from Bacillus cereus

greener alternative

≥200 units/mg protein

Synonym(s):

Lecithinase C, Lipophosphodiesterase I, PC-PLC, Phosphatidylcholine cholinephosphohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

lyophilized powder

Quality Level

specific activity

≥200 units/mg protein

mol wt

23-27 kDa by SDS-PAGE

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

solubility

H2O: soluble 1.0 mg/mL, faintly hazy to hazy

greener alternative category

storage temp.

−20°C

Gene Information

Bacillus cereus ATCC 10987 ... plC(2749087)

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in biodiesel research. For more information see the article in biofiles.

Application

Phospholipase C (PLC) is used to study adrenoceptor-mediated transmembrane signaling[1]. It is used to degrade inositol-containing phospholipids[2]. Phospholipase C from Bacillus, product P6621, has been used to study how flhF affects the export of PLC and other secretory virulence factors[3].
The enzyme from Sigma has been used in the digestion of phosphatidylserine fraction of Schistosoma mansoni adult worms.[4]

Biochem/physiol Actions

Hydrolyzes the phosphate bond on phosphatidylcholine and other glycerophospholipids yielding diacylglycerol; this enzyme will also hydrolyze the phosphate bonds of sphingomyelin, cardiolipin, choline plasmalogen and ceramide phospholipids.
Phospholipase C is induced by thrombin and platelet-activating factor, forming 1,2-diacylglycerol and phosphatidic acid[2]. PLC hydrolyzes the phosphate bond on phosphatidylcholine and other glycerophospholipids yielding diacylglycerol; this enzyme will also hydrolyze the phosphate bonds of sphingomyelin, cardiolipin, choline plasmalogen and ceramide phospholipids.

Unit Definition

One unit will liberate 1.0 μmole of water soluble organic phosphorus from L-α-phosphatidylcholine per min at pH 7.3 at 37 °C.

Physical form

Lyophilized powder containing approx. 10% protein. Remainder; trehalose, zinc sulfate, and potassium phosphate

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Desiree van der Kleij et al.
The Journal of biological chemistry, 277(50), 48122-48129 (2002-10-03)
Schistosome infections are characterized by prominent T cell hyporesponsiveness during the chronic stage of infection. We found that schistosome-specific phosphatidylserine (PS) activated TLR2 and affected dendritic cells such that mature dendritic cells gained the ability to induce the development of
W Siess et al.
The Journal of biological chemistry, 259(13), 8286-8292 (1984-07-10)
The present study compares the molecular mechanism by which thrombin, platelet-activating factor, and epinephrine induce platelet activation. Thrombin and platelet-activating factor induce an initial activation of phospholipase C, as measured by formation of 1,2-diacylglycerol and phosphatidic acid, during platelet shape
Clarisse Techer et al.
Food microbiology, 86, 103317-103317 (2019-11-11)
The famous French dessert "ile flottante" consists of a sweet egg white foam floating on a vanilla custard cream, which contains highly nutritive raw materials, including milk, sugar and egg. Spoilage issues are therefore a key concern for the manufacturers.
T Das et al.
The Journal of biological chemistry, 268(36), 27398-27405 (1993-12-25)
Our studies on the alpha 1-adrenoceptor signaling have demonstrated that the Gh family is a signal mediator. We report here that a 69-kDa phospholipase C (PLC) is the effector in this signal pathway. The enzyme was isolated by dissociating a
Sara Salvetti et al.
Microbiology (Reading, England), 153(Pt 8), 2541-2552 (2007-07-31)
Flagellar arrangement is a highly conserved feature within bacterial species. However, only a few genes regulating cell flagellation have been described in polar flagellate bacteria. This report demonstrates that the arrangement of flagella in the peritrichous flagellate Bacillus cereus is

Questions

  1. Could you please provide the recommended storage temperature for the solution of this product?

    1 answer
    1. The recommended storage for this product is at -20°C in working aliquots.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service