Skip to Content
Merck
All Photos(3)

Key Documents

P7633

Sigma-Aldrich

Phospholipase C from Clostridium perfringens (C. welchii)

greener alternative

Type I, lyophilized powder, 10-50 units/mg protein

Synonym(s):

Lecithinase C, Lipophosphodiesterase I, PC-PLC, Phosphatidylcholine cholinephosphohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial (Clostridium perfringens)

Quality Level

type

Type I

form

lyophilized powder

specific activity

10-50 units/mg protein

composition

protein, 20-40%

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

diagnostic assay manufacturing

greener alternative category

storage temp.

−20°C

Gene Information

Clostridium perfringens str. 13 ... plc(988262)

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in biodiesel research. For more information see the article in biofiles.

Application

Phospholipase C (PLC) has been used to study adrenoceptor-mediated transmembrane signaling. It is also used to degrade inositol-containing phospholipids. Furthermore, it has been used to study 9E3 gene activation.

Biochem/physiol Actions

Hydrolyzes the phosphate bond on phosphatidylcholine and other glycerophospholipids yielding diacylglycerol; this enzyme will also hydrolyze the phosphate bonds of sphingomyelin, cardiolipin, choline plasmalogen and ceramide phospholipids.
PLC hydrolyzes the phosphate bond on phosphatidylcholine and other glycerophospholipids yielding diacylglycerol. This enzyme also hydrolyzes the phosphate bonds of sphingomyelin, cardiolipin, choline plasmalogen and ceramide phospholipids. Phospholipase C is induced by thrombin and platelet-activating factor, forming 1,2-diacylglycerol and phosphatidic acid.

Unit Definition

One unit will liberate 1.0 μmole of water soluble organic phosphorus from egg yolk L-α-phosphatidylcholine per min at pH 7.3 at 37 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mojtaba Alimolaei et al.
Probiotics and antimicrobial proteins, 10(2), 251-257 (2017-04-13)
The alpha-toxin is one of the virulence factors of Clostridium perfringens for gas gangrene in humans and animals or necrotic enteritis in poultry. The C-terminal domain of this toxin ( cpa 247-370 ) was synthesized and cloned into pT1NX vector
R Spangler et al.
Proceedings of the National Academy of Sciences of the United States of America, 86(18), 7017-7021 (1989-09-01)
Induction of the transformation-related gene 9E3 by the v-src and v-fps gene products (v-Src and v-Fps) is blocked in chicken embryo fibroblasts depleted of protein kinase C (PKC). PKC agonists induce 9E3 gene expression. Protein kinase inhibitors block v-Src- and
Purification of Clostridium perfringens phospholipase C (alpha-toxin) by affinity chromatography on agarose-linked egg-yolk lipoprotein.
T Takahashi et al.
Biochimica et biophysica acta, 351(1), 155-171 (1974-05-10)
N Sieger et al.
Arthritis and rheumatism, 65(3), 770-779 (2012-12-13)
CD22 is a surface molecule exclusively expressed on B cells that regulates adhesion and B cell receptor (BCR) signaling as an inhibitory coreceptor of the BCR. Central downstream signaling molecules that are activated upon BCR engagement include spleen tyrosine kinase
Zahra Timsah et al.
Nature structural & molecular biology, 21(2), 180-188 (2014-01-21)
FGFR2-expressing human cancer cells with low concentrations of the adaptor protein Grb2 show high prevalence for metastatic outcome. In nonstimulated cells, the SH3 domain (and not the SH2 domains) of Plcγ1 directly competes for a binding site at the very

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service