Skip to Content
Merck
All Photos(2)

Key Documents

P8541

Sigma-Aldrich

Potassium sulfate

suitable for plant cell culture

Synonym(s):

K2SO4, sulphate of potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2SO4
CAS Number:
Molecular Weight:
174.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.71
Assay:
≥99% (titration)
form:
powder
solubility:
H2O: 10/150 g/mL, clear
Pricing and availability is not currently available.

Assay

≥99% (titration)

Quality Level

form

powder

technique(s)

cell culture | plant: suitable

solubility

H2O: 10/150 g/mL, clear

SMILES string

[K+].[K+].[O-]S([O-])(=O)=O

InChI

1S/2K.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

OTYBMLCTZGSZBG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashley C Parker et al.
Clinical orthopaedics and related research, 469(11), 3008-3015 (2011-05-10)
Local drug delivery has substantial potential to prevent infections compared with systemic delivery. Although calcium sulfate (CaSO(4)) has been studied for local drug delivery and two types are commercially available, it is unknown whether they differentially release antibiotics. We determined
Jinyong Yan et al.
Bioresource technology, 102(7), 4755-4758 (2011-02-01)
A dual modification procedure composed of cross-linking and protein coating with K(2)SO(4) was employed to modify Geotrichum sp. lipase for catalyzing biodiesel production from waste cooking oil. Compared to single modification of protein coating with K(2)SO(4), the dual modification of
C C Constantinoiu et al.
Poultry science, 86(9), 1910-1914 (2007-08-21)
Immunoglobulin Y is different from most of the other immunoglobulins because it does not bind protein A or protein G. Thiophilic gel chromatography has been successfully used to purify IgY from chicken egg yolk, but the technology has not previously
Jingjing Zheng et al.
Bioresource technology, 110, 224-231 (2012-02-15)
This study investigated the preparation and characteristics of protein-coated microcrystals (PCMCs) from Pseudomonas cepacia lipase (PS) and K(2)SO(4), and their application in biodiesel synthesis, via single factorial experiments and response surface methodology (RSM), the optimized PCMC-PS exhibited high activity and
Xinjian Shi et al.
Nature communications, 5, 4775-4775 (2014-09-03)
Tungsten trioxide/bismuth vanadate heterojunction is one of the best pairs for solar water splitting, but its photocurrent densities are insufficient. Here we investigate the advantages of using helical nanostructures in photoelectrochemical solar water splitting. A helical tungsten trioxide array is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service