Skip to Content
Merck
All Photos(3)

Documents

P7125

Sigma-Aldrich

Pepsin from porcine gastric mucosa

powder, ≥400 units/mg protein

Synonym(s):

Pepsin A, Pepsin from hog stomach

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
42010127
NACRES:
NA.54

biological source

Porcine gastric mucosa

form

powder

specific activity

≥400 units/mg protein

mol wt

35 kDa

solubility

10 mM HCl: soluble 1.0 mg/mL, clear to faintly turbid, colorless

UniProt accession no.

storage temp.

2-8°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Pepsin is a peptidase used to digest proteins and is commonly used in the preparation of Fab fragments from antibodies. Pepsin, from porcine gastric mucosa, has been used to hydrolyze dry cervical samples in mice. Product P7125 is provided in powder form. Product P7125 has been used to denature DNA from kidney cells and to digest pathology samples from anal canal carcinomas (ACC) biopsies prior to EGFR staining.
The enzyme from Sigma has been used to obtain total vitamin B12 content in food products prior using immunoaffinity columns It has also been used to digest minced soft tissue of snails prior to the isolation of the third stage larvae (L3).
Pepsin cleavage can be used to produce F(ab′)2 fragments of antibodies. pepsin at www.sigma-aldrich.com/enzymeexplorer.

Biochem/physiol Actions

It does not cleave at valine, alanine, or glycine linkages. Z-L-tyrosyl-L-phenylalanine, Z-L-glutamyl-L-tyrosine, or Z-L-methionyl-L-tyrosine may be used as substrates for pepsin digestion. Pepsin is inhibited by several phenylalanine-containing peptides.
Pepsin hydrolyzes peptide bonds, not amide or ester linkages. Pepsin cleaves peptides with an aromatic acid on either side of the peptide bond. Sulfur-containing amino acids increase susceptibility to hydrolysis when they are close to the peptide bond. Pepsin preferentially cleaves at the carboxyl side of phenylalanine and leucine and at the carboxyl side of glutamic acid residues. Cleaves Phe-Val, Gln-His, Glu-Ala, Ala-Leu, Leu-Tyr, Tyr-Leu, Gly-Phe, Phe-Phe and Phe-Tyr bonds in the β chain of insulin
Pepsin is the major proteolytic enzyme produced in the stomach. It digests proteins through the cleavage of interior peptide linkages.
Preferential cleavage: hydrophobic and aromatic residues in P1 and P1′ postitions. Cleaves Phe-Val, Gln-His, Glu-Ala, Ala-Leu, Leu-Tyr, Tyr-Leu, Gly-Phe, Phe-Phe and Phe-Tyr bonds in the β chain of insulin

Unit Definition

One unit will produce a ΔA280 of 0.001 per min at pH 2.0 at 37 °C, measured as TCA-soluble products using hemoglobin as substrate. (Final volume = 16 mL. Light path = 1 cm.)

Analysis Note

Optimum pH is 2-4. Active in 4 M urea and 3 M guanidine HCl. Stable at 60 °C. Pepsin is irreversibly inactivated at pH 8.0 - 8.5.
Protein determined by E1%/280

Other Notes

View more information on pepsin at www.sigma-aldrich.com/enzymeexplorer.

inhibitor

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Dahle et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(5), 1773-1778 (1997-03-04)
Microcolonies of 2-8 Madison-Darby canine kidney cells (MDCK II) and Chinese hamster lung fibroblasts (V79) cells were incubated with the photosensitizer Photofrin and exposed to light, and the resulting number of dead cells per colony was determined. The distribution of
Erli Neuhauss et al.
Memorias do Instituto Oswaldo Cruz, 102(1), 49-52 (2007-02-13)
Introduction of Achatina fulica in Brazil has led to serious concerns about its role as vector for metaIylid worms: AngioIylus costaricensis and A. cantonensis. Experimental infection with both parasites was performed to evaluate the potential risk for their transmission by
Lyly H Lê et al.
American journal of clinical pathology, 124(1), 20-23 (2005-06-01)
Most anal canal carcinomas (ACCs) are squamous cell carcinomas (SCCs). SCCs in other tumor sites strongly express epidermal growth factor receptors (EGFRs), the inhibition of which might result in favorable changes in tumor growth. A review of the published scientific
Xu-Dong Kong et al.
Nature biomedical engineering, 4(5), 560-571 (2020-05-13)
The oral administration of peptide drugs is hampered by their metabolic instability and limited intestinal uptake. Here, we describe a method for the generation of small target-specific peptides (less than 1,600 Da in size) that resist gastrointestinal proteases. By using
O Heudi et al.
Journal of chromatography. A, 1101(1-2), 63-68 (2005-10-12)
A new, faster and simple method to quantify Vitamin B12, both in foods and in premixes, by reversed-phase liquid chromatography with UV detection has been developed. Vitamin B12 was extracted from food products with 50 mM sodium acetate buffer pH

Protocols

This procedure may be used for determination of Pepsin activity using hemoglobin as the substrate. It is a spectrophotometric stop rate determination.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service