Skip to Content
Merck
All Photos(1)

Documents

77160

Sigma-Aldrich

Pepsin from porcine gastric mucosa

powder, slightly beige, ≥500 U/mg

Synonym(s):

Pepsin A, Pepsin from hog stomach

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
42010127
NACRES:
NA.54

form

powder

specific activity

≥500 U/mg

mol wt

35 kDa

color

slightly beige

UniProt accession no.

storage temp.

2-8°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Pepsin cleavage can be used to produce F(ab′)2 fragments of antibodies. pepsin at www.sigma-aldrich.com/enzymeexplorer.

Biochem/physiol Actions

Preferential cleavage: hydrophobic and aromatic residues in P1 and P1′ postitions. Cleaves Phe-Val, Gln-His, Glu-Ala, Ala-Leu, Leu-Tyr, Tyr-Leu, Gly-Phe, Phe-Phe and Phe-Tyr bonds in the β chain of insulin

Unit Definition

One unit corresponds to the amount of enzyme which increases the absorbance at 280 nm by 0.001 per minute at pH 2.0 and 37 °C (Hemoglobin, Catalog No. 51290, as substrate); 15,000 absorbance units as described above are equivalent to ∼1 Bergmeyer unit. One Bergmeyer unit is the amount of enzyme which hydrolyzes 1 μmole Acetyl-L-phenyl-3,5-diiodo-L-tyrosine at pH 2.0 and 37 °C.

Analysis Note

Optimum pH is 2-4. Active in 4 M urea and 3 M guanidine HCl. Stable at 60 °C. Pepsin is irreversibly inactivated at pH 8.0 - 8.5.

Other Notes

Peptide synthesis, Review.
Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.S. Fruton
Carlsberg Research Communications, 49, 41-41 (1984)
C A Abdel Malak et al.
International journal of peptide and protein research, 41(2), 97-101 (1993-02-01)
Swine pepsin at pH 5 efficiently catalyzes a condensation between Z-Ala-Ala-Phe-OH and p-nitroanilides of Leu, Phe, Val, Ala and Arg that leads to formation of corresponding benzyloxycarbonyl-tetrapeptide p-nitroanilides with yields of 70-90%. These reactions are complicated by co-precipitation of pepsin
Ludovic Hermabessiere et al.
The Science of the total environment, 749, 141651-141651 (2020-08-25)
Plastic pollution is a source of chemical to the environment and wildlife. Despite the ubiquity of plastic pollution and thus plastic additive in the environment, plastic additives have been studied to a limited extend. As a prerequisite to a study
M.P. Bemquerer et al.
Biomedica Biochimica Acta, 50 (1991)
Abdelhacib Kihal et al.
Animals : an open access journal from MDPI, 11(8) (2021-08-28)
The aim of this study was to determine the capacity of six mycotoxin binders (MTBs) to adsorb vitamins A, D and E in an in vitro system that simulates gastric and intestinal digestion. Experiment 1 evaluated the recovery rate of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service