Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pepsin from porcine gastric mucosa
Synonym(s):
Pepsin A, Pepsin from hog stomach
CAS Number:
9001-75-6
EC Number:
232-629-3
Products
Product Category
proteases (8)
Brand
Sigma-Aldrich (8)
Biological Source
Color
Physical Form
Agency
Tissue
Quality Segment
Application
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
P7000
Pepsin from porcine gastric mucosa,
powder, ≥250 units/mg solid
Expand
View Pricing
P7012
Pepsin from porcine gastric mucosa,
lyophilized powder, ≥2,500 units/mg protein (E1%/280)
Expand
View Pricing
P6887
Pepsin from porcine gastric mucosa,
lyophilized powder, ≥3,200 units/mg protein
Expand
View Pricing
P7125
Pepsin from porcine gastric mucosa,
powder, ≥400 units/mg protein
Expand
View Pricing
77160
Pepsin from porcine gastric mucosa,
powder, slightly beige, ≥500 U/mg
Expand
View Pricing
77151
Pepsin from porcine gastric mucosa,
powder, slightly beige, 1200-2400 U/mg
Expand
View Pricing
SRE0001
Pepsin from porcine gastric mucosa,
Suitable for manufacturing of diagnostic kits and reagents, lyophilized powder, ≥3200 units/mg protein
Expand
View Pricing
77161
Pepsin from porcine gastric mucosa,
tested according to Ph. Eur.
Expand
View Pricing