Skip to Content
Merck
All Photos(1)

Documents

P4739

Sigma-Aldrich

Phosphate buffered saline

powder (dry milled), pH 7.3, with 5% nonfat milk

Synonym(s):

PBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161503
NACRES:
NA.25

product name

Phosphate buffered saline, with 5% nonfat milk, powder (dry milled), pH 7.3

form

powder (dry milled)

composition

nonfat dry milk
potassium chloride, 76.5%
potassium phosphate monobasic anhydrous, 0.3%
sodium chloride, 12.2%
sodium phosphate dibasic, 10.7%

pH

7.3

Looking for similar products? Visit Product Comparison Guide

Application

Phosphate-buffered saline has been used to block the SARS-COV2 antigen during the preparation of ELISA plates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

he Protease Inhibitor 4-(2-Aminoethyl) Benzenesulfonyl Fluoride Covalently Modifies Human Plasma Proteins and Changes Protein Isoform Profiles
Mark Schuchard
Molecular and Cellular Proteomics, 3 (2004)
Xiao Cui et al.
Cell biology international, 44(9), 1811-1819 (2020-05-07)
Multiple sclerosis (MS) is an autoimmune disease of the central nervous system that is caused by the interaction of genetic and environmental factors. Current studies have shown that Fc-receptor like-3 (FCRL3) is closely related to MS, but the specific role
Campylobacter jejuni in penguins, Antarctica.
Petra Griekspoor et al.
Emerging infectious diseases, 15(5), 847-848 (2009-05-01)
Yong N Li et al.
The Journal of comparative neurology, 516(5), 442-453 (2009-08-06)
Horizontal cells (HCs) are involved in establishing the center-surround receptive field organization of photoreceptor and bipolar cells. In many species, HCs respond differentially to colors and may play a role in color vision. An earlier study from our laboratory suggested
Konrad Franke et al.
Translational oncology, 2(4), 310-320 (2009-12-04)
Nodal spread is the single most important prognostic factor of survival in gastric cancer patients. In this study, genes that were upregulated in the lymph node metastases of gastric cancer were identified and may serve as putative novel therapeutic target.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service