Skip to Content
Merck
All Photos(1)

Key Documents

P4458

Sigma-Aldrich

Penicillin-Streptomycin

liquid, non-animal origin, suitable for cell culture, BioReagent

Synonym(s):

Pen-Strep

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.76

Product Name

Penicillin-Streptomycin, Solution Stabilized, with 5,000 units penicillin and 5mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture

Quality Level

sterility

0.1 μm filtered

product line

BioReagent

form

liquid

concentration

50 ×

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes
protein synthesis | interferes

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC1(C)S[C@@H]2[C@H](NC(=O)Cc3ccccc3)C(=O)N2[C@H]1C(O)=O.CNC[C@H]4[C@H](O)[C@@H](O)[C@H](CCO)O[C@H]4O[C@H]5[C@@H](O[C@@H](C)C5(O)C=O)O[C@H]6[C@H](O)[C@@H](O)[C@H](NC(N)=N)[C@@H](O)[C@@H]6NC(N)=N

InChI

1S/C23H43N7O12.C16H18N2O4S/c1-7-23(38,6-32)18(42-19-8(5-28-2)12(33)13(34)9(40-19)3-4-31)20(39-7)41-17-11(30-22(26)27)14(35)10(29-21(24)25)15(36)16(17)37;1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9/h6-20,28,31,33-38H,3-5H2,1-2H3,(H4,24,25,29)(H4,26,27,30);3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22)/t7-,8-,9-,10+,11-,12-,13-,14+,15-,16+,17+,18-,19-,20-,23+;11-,12+,14-/m01/s1

InChI key

KMOOCZWLFBSQCW-XFOLRDBHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Penicillin, a β-lactam antibiotic is extremely active towards bacteria. Penicillins are also termed as penams. It is naturally occurring and has a bicyclic skeleton. Streptomycin is a pseudotrisaccharide. It comprises of a monosubstituted aminocyclitol.

Application

Penicillin-Streptomycin has been used:
  • as a supplement in Dulbecco′s modified Eagle′s medium (DMEM) to maintain the Huh7 liver hepatoma cell line
  • as a supplement in Roswell park memorial institute-1640 (RPMI) medium to maintain mouse lymphoma YAC-1 cells
  • as a component in minimum essential medium Eagle to treat individual mouse spleens for the preparation of splenocytes
  • as a supplement in FMX-8 complete medium to culture Chinese hamster ovary cells

Recommended for use in cell culture media at 20 ml/L.

Biochem/physiol Actions

Penicillins can prevent the synthesis of bacterial cell wall. This antibiotic can be used in the treatment of skin and urinary tract infections, upper and lower respiratory infections, and endocarditis. Streptomycin can block Mycobacterium tuberculosis in vitro. Streptomycin inhibits prokaryote protein synthesis by preventing the transition from initiation complex to chain-elongating ribosome, causing miscoding.

Components

This product is formulated to contain 5,000 units penicillin and 5 mg streptomycin/mL solubilized in a proprietary citrate buffer.

Other Notes

20ml,100ml
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Penicillins
xPharm: The Comprehensive Pharmacology Reference (2016)
Effect of Lactobacillus brevis KB290 on the cell-mediated cytotoxic activity of mouse splenocytes: a DNA microarray analysis
Fukui Y, et al.
The British Journal of Nutrition, 110(9), 1617-1629 (2013)
Aminoglycosides Antibiotics
Comprehensive Medicinal Chemistry II, 7, 629-652 (2007)
Streptomyces-derived quorum-sensing systems engineered for adjustable transgene expression in mammalian cells and mice
Weber W, et al.
Nucleic Acids Research, 31(14), e71-e71 (2003)
David Roquis et al.
PLoS pathogens, 14(5), e1007066-e1007066 (2018-05-22)
Epigenetic mechanisms and chromatin structure play an important role in development. Their impact is therefore expected to be strong in parasites with complex life cycles and multiple, strikingly different, developmental stages, i.e. developmental plasticity. Some studies have already described how

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service