Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Penicillin-Streptomycin
Products
Product Category
antibiotics (4)
Brand
Sigma-Aldrich (3)
Manufacturer
Physical Form
Product Line
Activity Spectrum
Quality Segment
Mode of Action
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
P4333
Penicillin-Streptomycin,
liquid, non-animal origin, suitable for cell culture, BioReagent
Expand
View Pricing
P0781
Penicillin-Streptomycin,
liquid, non-animal origin, suitable for cell culture, BioReagent
Expand
View Pricing
P4458
Penicillin-Streptomycin,
liquid, non-animal origin, suitable for cell culture, BioReagent
Expand
View Pricing
11074440001
Penicillin-Streptomycin,
suitable for cell culture, lyophilized (clear, colorless solution after reconstitution), non-sterile; 0.2 μm filtered, sufficient for 20 mL
Expand
View Pricing