Skip to Content
Merck
All Photos(6)

Key Documents

P3088

Sigma-Aldrich

Anti-Parvalbumin Antibody

mouse monoclonal, PARV-19

Synonym(s):

Parvalbumin Antibody, Parvalbumin Antibody - Monoclonal Anti-Parvalbumin antibody produced in mouse

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

Product Name

Monoclonal Anti-Parvalbumin antibody produced in mouse, clone PARV-19, ascites fluid

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

PARV-19, monoclonal

mol wt

antigen 12 kDa

contains

15 mM sodium azide

species reactivity

fish, frog, rabbit, canine, feline, bovine, pig, human, goat, rat

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable (10-20K titer using Mouse skeletal muscle extract expressing Parvalbumin protein)

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PVALB(5816)
rat ... Pvalb(25269)

General description

PVALB (parvalbumin) is a classical small member of the low molecular weight, acidic protein family known as calcium binding proteins (CaBPs) EF-hand superfamily. This protein shows a restricted pattern of expression in brain and muscle. It is a cytoplasmic high affinity CaBP.
Monoclonal Anti-Parvalbumin (mouse IgG1 isotype) is derived from the PARV-19 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Parvalbumin is a calcium-binding protein.

Specificity

Recognizes parvalbumin in a Ca2+ ion-dependent manner. Does not react with other members of the EF-hand family such as calmodulin, intestinal calcium-binding protein, S100A2 (S100L), S100A6 (calcyclin), the α chain of S-100 (i.e. in S-100a and S-100ao), or the β chain (i.e. in S-100a and S-100b).

Immunogen

Frog muscle parvalbumin.

Application

Monoclonal Anti-Parvalbumin antibody produced in mouse has been used for immunohistochemistry, immunocytochemistry, and immunofluorescence.

Biochem/physiol Actions

PVALB (parvalbumin) protein modulates relaxation in fast contracting muscles, where it localizes to type II (fast-twitch) mammalian skeletal muscle fibres in small mammals. This protein is a crucial regulator of neuronal intracellular calcium dynamics. PVALB protein maintains synaptic transmission during bursts, where it prevents presynaptic Ca2+ accumulation and hence the harmful effects of excess presynaptic Ca2+ levels. It also maintains synaptic transmission by allowing for linear information processing post-synaptically by preventing cumulative neuronal facilitation.

PVALB (parvalbumin) expression serves as marker for differentiating tumors as benign and malignant, in relation with thyroid lesions. Decreased expression of parvalbumin positive interneurons in rat model is associated with schizophrenia, and is known to disturb neuronal activity.

Physical form

Liquid

Storage and Stability

For continuous use, store at 2-8 °C for up to one month.
For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aging-related changes in calcium-binding proteins in rat perirhinal cortex
Moyer Jr J R, et al.
Neurobiology of Aging, 32(9), 1693-1706 (2011)
Age-dependent in vivo conversion of mouse cochlear pillar and Deiters' cells to immature hair cells by Atoh1 ectopic expression.
Liu Z et al
The Journal of Neuroscience, 32(19), 6600-6610 (2012)
PVALB diminishes [Ca2+] and alters mitochondrial features in follicular thyroid carcinoma cells through AKT/GSK3β pathway}
Mendes T B, et al.
Endocrine-Related Cancer, 23(9), 769-782 (2016)
A loss of parvalbumin-containing interneurons is associated with diminished oscillatory activity in an animal model of schizophrenia.
Lodge DJ, et al.
The Journal of Neuroscience, 29(8), 2344-2354 (2009)
Role of the calcium-binding protein parvalbumin in short-term synaptic plasticity.
Caillard O et al
Proceedings of the National Academy of Sciences of the USA, 97(24), 13372-13377 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service