Skip to Content
Merck
All Photos(4)

Key Documents

C8035

Sigma-Aldrich

Anti-Aggrecan Antibody

Monoclonal Anti-Chondroitin Sulfate antibody produced in mouse

mouse monoclonal, CS-56

Synonym(s):

Chondroitin Sulfate Antibody, Chondroitin Sulfate Antibody - Monoclonal Anti-Chondroitin Sulfate antibody produced in mouse, Cs56 Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Monoclonal Anti-Chondroitin Sulfate antibody produced in mouse, clone CS-56, ascites fluid

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

CS-56, monoclonal

species reactivity

chicken, bovine, mouse, human, pig, rabbit, rat

technique(s)

indirect immunofluorescence: 1:200 using bovine mammary gland epithelial (BMGE) cells
microarray: suitable

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACAN(176)
mouse ... Acan(11595)
rat ... Acan(58968)

General description

Chondroitin sulfate is a linear polysaccharide composed of alternating disaccharide glucuronic acid and N-acetylgalactosamine. It contains proteoglycan (PG) of the extracellular matrix (ECM). Monoclonal Anti-Chondroitin Sulfate (mouse IgM isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.
This product can be found as purified product that was produced using cell culture hybridoma (SAB4200696)

Specificity

The antibody has been reported to be specific for the glycosaminoglycan (GAG) portion of native chondroitin sulfate proteoglycan (CSPG). The antibody reacts specifically with chondroitin sulfate types A and C but not with type B (dermatan sulfate), and may be used for localization of chondroitin sulfate in tissues and cultured fibroblasts.

Immunogen

Ventral membranes of chicken gizzard fibroblasts

Application

Monoclonal Anti-Chondroitin Sulfate antibody produced in mouse was used:
  • in immunohistochemistry as primary antibody to study of spinal cords and brain cryosections.
  • in ELISA and western blot for purification of soluble forms of myelin-associated glycoprotein (MAG) and neural cell adhesion molecule (N-CAM).
  • in flow cytometry studies.
  • as primary antisera in a study of TGF-β regulation.
Monoclonal Anti-Chondroitin Sulfate can also be used to study chondroitin sulfate proteoglycan (CSPG) distribution and its relationships to specific cell-substrate contacts. It is also suitable for microarray and indirect immunofluorescence at the dilution of 1:200 by using bovine mammary gland epithelial (BMGE) cells.

Biochem/physiol Actions

Chondroitin sulfate is involved in tumor invasion and also plays a vital role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes. In Chondroitin sulfate proteoglycan (CSPG), proteoglycan part is involved in cell proliferation and migration which stimulates endothelial cells motility during microvascular morphogenesis. Chondroitin sulfate is used in osteoarthritis treatment. Upregulation of chondroitin sulfate proteoglycans is observed after neuronal injury. It possess the ability to inhibit axonal regeneration.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month.
For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Other Notes

This product can be found as purified product that was produced using cell culture hybridoma product.
SAB4200696 Anti-Chondroitin Sulfate antibody, Mouse monoclonal
clone CS-56, purified from hybridoma cell culture

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PTPσ is a receptor for chondroitin sulfate proteoglycan, an inhibitor of neural regeneration.
Shen Y, et al.
Science, 326(5952) (2009)
Effect of the counterion behavior on the frictional?compressive properties of chondroitin sulfate solutions
Polymer, 50(7), 1805-1813 (2009)
Charlotte Rolny et al.
The Journal of biological chemistry, 277(22), 19315-19321 (2002-03-26)
Platelet-derived growth factor (PDGF) induces mitogenic and migratory responses in a wide variety of cells, by activating specific receptor tyrosine kinases denoted the PDGF alpha- and beta-receptors. Different PDGF isoforms bind in a distinct manner to glycosaminoglycans, particularly heparan sulfate.
M Sobue et al.
Stain technology, 64(1), 43-47 (1989-01-01)
Dermatan sulfate proteoglycan chains were detected in tissue sections treated with chondroitin B-lyase (0.01 units/ml) in 20 mM Tris-HCl (pH 8.0) for 1 hr, followed by staining with antibody 9A2 specific for unsaturated uronic acid coupled to N-acetylgalactosamine-4 sulfate. In
Z Avnur et al.
Cell, 38(3), 811-822 (1984-10-01)
Chondroitin-sulfate containing proteoglycan (CSPG) of the extracellular matrix (ECM) was visualized in chick tissues and cell cultures with a monoclonal antibody, CS-56. Cultured cells of various origins contained dense punctate layers of CSPG on both the substrate and the cell

Articles

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service