Skip to Content
Merck
All Photos(5)

Key Documents

MAB5406

Sigma-Aldrich

Anti-GAD1(GAD67) Antibody

CHEMICON®, mouse monoclonal, 1G10.2

Synonym(s):

Glutamate Decarboxylase 67kDa Isoform, Glutamate Decarboxylase 1, Glutamic acid decarboxylase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1G10.2, monoclonal

species reactivity

human, mouse, rat

packaging

antibody small pack of 25 μg

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... GAD1(2571)

General description

Gutamic acid decarboxylase (GAD; E.C. 4.1.1.15) is the enzyme responsible for the conversion of glutamic acid to gamma-aminobutyric acid (GABA), the major inhibitory transmitter in higher brain regions, and putative paracrine hormone in pancreatic islets. Two molecular forms of GAD (65 kDa and 67 kDa, 64% aa identity between forms) are highly conserved and both forms are expressed in the CNS, pancreatic islet cells, testis, oviduct and ovary. The isoforms are regionally distributed cytoplasmically in the brains of rats and mice (Sheikh, 1999). GAD65 is an amphiphilic, membrane-anchored protein (585 a.a.), encoded on human chromosome 10, and is responsible for vesicular GABA production. GAD67 is cytoplasmic (594 a.a.), encoded on chromosome 2, and seems to be responsible for significant cytoplasmic GABA production. GAD expression changes during neural development in rat spinal cord. GAD65 is expressed transiently in commissural axons around E13 but is down regulated the next day while GAD67 expression increases mostly in the somata of those neurons (Phelps, 1999). In mature rat pancreas, GAD65 and GAD67 appear to be differentially localized, GAD65 primarily in insulin-containing beta cells and GAD67 in glucagon-containing (A) cells (Li, 1995). GAD67 expression seems to be particularly plastic and can change in response to experimental manipulation (for example neuronal stimulation or transection) or disease progression and emergent disorders like schizophrenia (Volk, 2000). Colocalization of the two GAD isoforms also shows changes in GAD65/GAD67 distributions correlated with certain disease states such as IDDM and SMS.

Specificity

Anti-GAD67 Antibody, clone 1G10.2 reacts with the 67kDa isoform of Glutamate Decarboxylase (GAD67) of rat, mouse and human origins, other species not yet tested. No detectable cross-reactivity with GAD65 by Western blot on rat brain lysate when compared to blot probed with AB1511 that reacts with both GAD65 & GAD67.

Immunogen

Recombinant GAD67 protein

Application

Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neuronal & Glial Markers
This Anti-GAD67 Antibody, clone 1G10.2 is validated for use in IH, IH(P), WB for the detection of GAD67. This Anti GAD antibody has more than 75 product citations.
Western Blot: A 1:5,000-1:10,000 dilution was used on a previous lot using ECL on rat and mouse brain lysate.

Immunohistochemistry on 4% paraformaldehyde fixed mouse and rat tissue: 1:1,000-1:5,000. The antibody has also worked well on human tissue fixed for 10 minutes in 4% PFA at 4°C or 100% ethanol at +20°C or 100% acetone at -20°C at dilutions of 1:2,500-1:5,000.
A polyclonal to GAD67 is also available as AB5992.

Optimal working dilutions must be determined by end user.

Quality

Routinely evaluated by immunohistochemistry by SKNSH cell lysate.

Immunohistochemistry(paraffin) Analysis:
Representative staining pattern and morphology of GAD67 in somatosensory 1, barrel field of the mouse cerebral cortex. All brown spots are Lamina VI a neurons. No Epitope retrieval was necessary. This lot of the antibody was diluted to 1:500, using IHC Select Detection with HRP-DAB.
Optimal Staining pattern/morphology of GAD67: Mouse Brain.

Target description

67 kDa

Linkage

Replaces: AB5992; AB5862P

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG2a in buffer containing 0.1% sodium azide

Storage and Stability

Stable for 6 months at 2-8ºC in undiluted aliquots from date of receipt.

Analysis Note

Control
SKNSH cell lysate (human neuroblastoma), mouse cerebral cortex.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deletion of GAD67 in dopamine receptor-1 expressing cells causes specific motor deficits.
Heusner, CL; Beutler, LR; Houser, CR; Palmiter, RD
Genesis (2000)
Fu-Quan Huo et al.
Cerebral cortex (New York, N.Y. : 1991), 19(6), 1263-1272 (2008-11-05)
The ventrolateral orbital cortex (VLO) is part of an endogenous analgesic system, consisting of the spinal cord-thalamic nucleus submedius-VLO periaqueductal gray (PAG)-spinal cord loop. The present study examined morphological connections of GABAergic (gamma-aminobutyric acidergic) neurons and serotonergic projection terminals from
Hanno S Meyer et al.
Cerebral cortex (New York, N.Y. : 1991), 20(10), 2277-2286 (2010-06-11)
This is the second article in a series of three studies that investigate the anatomical determinants of thalamocortical (TC) input to excitatory neurons in a cortical column of rat primary somatosensory cortex (S1). Here, we report the number and distribution
Temporal patterns of fos expression in the dentate gyrus after spontaneous seizures in a mouse model of temporal lobe epilepsy.
Peng, Z; Houser, CR
The Journal of Neuroscience null
B Fritsch et al.
Neuroscience, 163(1), 415-429 (2009-06-23)
An acute brain insult such as traumatic head/brain injury, stroke, or an episode of status epilepticus can trigger epileptogenesis, which, after a latent, seizure-free period, leads to epilepsy. The discovery of effective pharmacological interventions that can prevent the development of

Questions

1–2 of 2 Questions  
  1. What is the volume in the 25ug vial?

    1 answer
    1. This product is a 1 mg/mL preparation. The 25 ug vial has a volume of 25 uL.

      Helpful?

  2. HI THERE IS THE CATALOG NUMBER MAB5406 but the ID is AB_2278725 is that right

    1 answer
    1. That is correct. MAB5406 is the product or catalog number assigned to the antibody. The AB_2278725 is the code assigned by the RRID. The product is cited in the Research Resource Identification Portal and the Antibody Registry and refers to the same item.
      Please see the link below to review the listing at the Antibody Registry:
      https://www.antibodyregistry.org/AB_2278725

      Please see the link below to review the RRID entry:
      https://scicrunch.org/resources/data/record/nif-0000-07730-1/AB_2278725/resolver?q=AB_2278725&l=AB_2278725&i=rrid:ab_2278725-67997

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service