Skip to Content
Merck
All Photos(1)

Key Documents

P1999

Sigma-Aldrich

PEAQX tetrasodium hydrate

≥98% (HPLC)

Synonym(s):

NVP-AAM077 tetrasodium hydrate, [[[(1S)-1-(4-Bromophenyl)ethyl]amino](1,2,3,4-tetrahydro-2,3-dioxo-5-quinoxalinyl)methyl] phosphonic acid tetrasodium hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13BrN3Na4O5P · xH2O
CAS Number:
Molecular Weight:
542.14 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

water: 10 mg/mL

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[H]O[H].C[C@H](NC(c1cccc2nc([O-])c([O-])nc12)P([O-])([O-])=O)c3ccc(Br)cc3

InChI

1S/C17H17BrN3O5P.4Na.H2O/c1-9(10-5-7-11(18)8-6-10)19-17(27(24,25)26)12-3-2-4-13-14(12)21-16(23)15(22)20-13;;;;;/h2-9,17,19H,1H3,(H,20,22)(H,21,23)(H2,24,25,26);;;;;1H2/q;4*+1;/p-4/t9-,17?;;;;;/m0...../s1

InChI key

SMGAGBKXHAHCGQ-VSYRWHDMSA-J

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
ACR5000PGA8971MSP1L
suitability

suitable for mass spectrometry

suitability

-

suitability

-

suitability

-

shipped in

wet ice

shipped in

wet ice

shipped in

-

shipped in

-

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

−20°C

Application

PEAQX tetrasodium hydrate has been used N-methyl-D-aspartate (NMDA) receptor 2A (NR2A) antagonist in rats[1][2] and mice.[3]

Biochem/physiol Actions

PEAQX is more specific for N-methyl-D-aspartate (NMDA) receptor 2A (NR2A) than N-methyl-D-aspartate (NMDA) receptor 2B (NR2B) and induces striatal apoptosis. It is less potent in activating caspase-3 than its stereomer NVP-AAM007.[4]
NMDA receptor antagonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Customers Also Viewed

    Slide 1 of 1

    1 of 1

    Melissa Morales et al.
    Behavioural brain research, 250, 18-22 (2013-05-09)
    Adolescents display high levels of interactions with peers relative to other age groups, with these interactions further enhanced by ethanol under some circumstances. Understanding of the neural mechanisms underlying these high levels of social interactions is important given that alcohol
    Melissa Morales et al.
    Psychopharmacology, 231(8), 1797-1807 (2013-09-18)
    NMDA antagonists consistently produce social inhibition in adult animals, although effects of these manipulations on social behavior of adolescents are relatively unknown. The aim of this study was to assess potential age differences in the socially inhibitory effects of the
    Yves P Auberson et al.
    Bioorganic & medicinal chemistry letters, 12(7), 1099-1102 (2002-03-23)
    NMDA antagonists derived from 5-phosphonomethyl-1,4-dihydroquinoxaline-2,3-dione (3a) are potent anticonvulsant agents, and display strong protective effects in the electroshock-induced convulsion assay in mice. Their preference for the human NMDAR 1A/2A over 1A/2B subunit composition was optimized, leading to (1RS,1'S)-PEAQX (9r), which
    N C Anastasio et al.
    Neuroscience, 163(4), 1181-1191 (2009-08-06)
    The mechanism underlying phencyclidine (PCP)-induced apoptosis in perinatal rats and the development of schizophrenia-like behaviors is incompletely understood. We used antagonists for N-methyl-D-aspartate (NMDA) receptor subunit NR2A- and NR2B-containing NMDA receptor to test the hypothesis that the behavioral and apoptotic
    Da Song et al.
    Neurobiology of learning and memory, 155, 435-445 (2018-09-24)
    The CA3 subregion of the hippocampus is important for rapid encoding, storage and retrieval of associative memories. Lesions and pharmacological inhibitions of hippocampal CA3 suggest that it is essential for different memories. However, how CA3 functions in spatial and episodic

    Articles

    We offers many products related to ion channel family glutamate receptors for your research needs.

    We offers many products related to ion channel family glutamate receptors for your research needs.

    We offers many products related to ion channel family glutamate receptors for your research needs.

    We offers many products related to ion channel family glutamate receptors for your research needs.

    See All

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service