Skip to Content
Merck
All Photos(4)

Documents

M8750

Sigma-Aldrich

(+)-Methamphetamine hydrochloride

Synonym(s):

d-Desoxyephedrine hydrochloride, d-N,α-Dimethylphenethylamine hydrochloride, Methylamphetamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N · HCl
CAS Number:
Molecular Weight:
185.69
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

H2O: soluble
ethanol: soluble

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

originator

Teva

SMILES string

Cl[H].CN[C@@H](C)Cc1ccccc1

InChI

1S/C10H15N.ClH/c1-9(11-2)8-10-6-4-3-5-7-10;/h3-7,9,11H,8H2,1-2H3;1H/t9-;/m0./s1

InChI key

TWXDDNPPQUTEOV-FVGYRXGTSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Sympathomimetic with more potent central effects than amphetamine. It is transported into terminals via the monoamine transporters and induces release of dopamine, norepinephrine, epinephrine, and serotonin. Dopamine release is important for the addictive properties of methamphetamine while norepinephrine and epinephrine release are important for the cardiovascular effects. Serotonin neurotoxin.

Features and Benefits

This compound was developed by Teva. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaveh Shahveisi et al.
Pharmacology, biochemistry, and behavior, 185, 172759-172759 (2019-08-16)
Susceptibility to interference can be a result of memory retrieval and reconsolidation. Given the fact that addiction develops through the neural mechanisms of learning and memory, it would not be surprising that a consolidated drug reward memory may also be
Noémie Cresto et al.
International journal of molecular sciences, 22(13) (2021-07-03)
Alpha-synuclein (α-syn) and leucine-rich repeat kinase 2 (LRRK2) play crucial roles in Parkinson's disease (PD). They may functionally interact to induce the degeneration of dopaminergic (DA) neurons via mechanisms that are not yet fully understood. We previously showed that the
Leila Rezaeian et al.
Iranian journal of basic medical sciences, 23(11), 1480-1488 (2020-11-26)
This research aimed at evaluating the effect of berberine hydrochloride on anxiety-related behaviors induced by methamphetamine (METH) in rats, assessing relapse and neuroprotective effects. 27 male Wistar rats were randomly assigned into groups of Control, METH-withdrawal (METH addiction and subsequent
G J Rivière et al.
The Journal of pharmacology and experimental therapeutics, 291(3), 1220-1226 (1999-11-24)
The purpose of these studies was to better understand the behavioral effects and pharmacokinetics of an i.v. bolus dose of (+)-methamphetamine [(+)-METH] in a rat model of (+)-METH abuse. We characterized the behavioral effects after increasing (+)-METH doses (0.1, 0.3
Arkadiusz Liśkiewicz et al.
Brain, behavior, and immunity, 80, 247-254 (2019-03-20)
Methamphetamine (METH) abusers are prone to develop a variety of comorbidities, including cognitive disabilities, and the immunological responses have been recognized as an important component involved in the toxicity of this drug. Cytokines are among the key mediators between systemic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service