Skip to Content
Merck
All Photos(1)

Documents

N6287

Sigma-Aldrich

Nutlin-3

≥98% (HPLC), powder, Mdm2 antagonist

Synonym(s):

(±)-4-[4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxy-phenyl)-4,5-dihydro-imidazole-1-carbonyl]-piperazin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H30Cl2N4O4
CAS Number:
Molecular Weight:
581.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Nutlin-3, ≥98% (HPLC), powder

ligand

nutlin-3

Assay

≥98% (HPLC)

form

powder

reaction suitability

reagent type: ligand

solubility

DMSO: 20 mg/mL
H2O: insoluble

originator

Roche

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C(N1CC(NCC1)=O)N2C(C3=CC=C(Cl)C=C3)C(C4=CC=C(Cl)C=C4)N=C2C5=C(OC(C)C)C=C(OC)C=C5

InChI

1S/C30H30Cl2N4O4/c1-18(2)40-25-16-23(39-3)12-13-24(25)29-34-27(19-4-8-21(31)9-5-19)28(20-6-10-22(32)11-7-20)36(29)30(38)35-15-14-33-26(37)17-35/h4-13,16,18,27-28H,14-15,17H2,1-3H3,(H,33,37)/t27-,28+/m1/s1

InChI key

BDUHCSBCVGXTJM-IZLXSDGUSA-N

Application

Nutlin-3 has been used:
  • as a drug to stimulate p53 functions in gene transfer experiment
  • to inject worms to verify the prevalent role of translationally controlled tumor protein (TCTP) in posterior amputated Eeugeniae
  • as a p53 activator in cyclotherapy studies
  • as an mdm2 inhibitor to know its effect on p53 levels, cleaved caspase 3 and Poly (ADP-ribose) polymerase (PARP) cleavage

Biochem/physiol Actions

Nutlin-3, an antagonist of a human homolog of murine double minute 2 (HDM2). It has the ability to inhibit the HDM2-directed degradation of p53. Nutlin-3 can also enhance the transcriptional activities of p73.
Nutlin-3 is a Mdm2 (mouse double minute 2) antagonist, p53 pathway activator, and apoptosis inducer.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Other Notes

Relative stereochemistry; racemic mixture of (4R, 5S) and (4S, 5R) enantiomers.

Analysis Note

relative stereochemistry

Legal Information

Sold under license from Hoffman-La Roche, Inc. US patent 6,734,302.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amine Bouafia et al.
PLoS genetics, 10(5), e1004309-e1004309 (2014-05-17)
Genomic instability is a major hallmark of cancer. To maintain genomic integrity, cells are equipped with dedicated sensors to monitor DNA repair or to force damaged cells into death programs. The tumor suppressor p53 is central in this process. Here
HDM2 antagonist Nutlin-3 disrupts p73-HDM2 binding and enhances p73 function
Lau LMS, et al.
Oncogene, 27(7), 997-997 (2008)
Activation of endogenous p53 by combined p19Arf gene transfer and nutlin-3 drug treatment modalities in the murine cell lines B16 and C6
Merkel CA, et al.
BMC Cancer, 10(1), 316-316 (2010)
Function of translationally controlled tumor protein (TCTP) in Eudrilus eugeniae regeneration
Subramanian ER, et al.
PLoS ONE, 12(4), e0175319-e0175319 (2017)
Makoto Miyazaki et al.
Cell death discovery, 4, 56-56 (2018-05-16)
Anaplastic lymphoma kinase (ALK) is an oncogenic receptor tyrosine kinase that is activated by gene amplification and mutation in neuroblastomas. ALK inhibitors can delay the progression of ALK-driven cancers, but are of limited use owing to ALK inhibitor resistance. Here

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service