Skip to Content
Merck
All Photos(1)

Key Documents

T2320

Sigma-Aldrich

T0901317

≥98%

Synonym(s):

N-(2,2,2-Trifluoroethyl)-N-[4-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl]phenyl]-benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12NSO3F9
CAS Number:
Molecular Weight:
481.33
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98%

mol wt

481.3

storage temp.

−20°C

SMILES string

OC(c1ccc(cc1)N(CC(F)(F)F)S(=O)(=O)c2ccccc2)(C(F)(F)F)C(F)(F)F

InChI

1S/C17H12F9NO3S/c18-14(19,20)10-27(31(29,30)13-4-2-1-3-5-13)12-8-6-11(7-9-12)15(28,16(21,22)23)17(24,25)26/h1-9,28H,10H2

InChI key

SGIWFELWJPNFDH-UHFFFAOYSA-N

Gene Information

Related Categories

Application

T0901317 has been used in LXR agonist study in order to observe the expression of all dietary cholesterol-regulated genes in mice. It has also been used to stimulate the radiolabeled peritoneal macrophages from WT (wildtype) and Tg (transgenic) mice for cellular cholesterol efflux assay.

Biochem/physiol Actions

LXR agonist whose treatment results in an LXR-dependent up-regulation of ABC1 gene expression
T0901317 can prevent the secretion of insulin by acting on the metabolism of mitochondria.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The LXR ligand T0901317 acutely inhibits insulin secretion by affecting mitochondrial metabolism
Maczewsky J, et al.
Endocrinology, 158(7), 2145-2154 (2017)
Differential Gene Regulation of StarD4 and StarD5 Cholesterol Transfer Proteins ACTIVATION OF StarD4 BY STEROL REGULATORY ELEMENT-BINDING PROTEIN-2 AND StarD5 BY ENDOPLASMIC RETICULUM STRESS
Soccio RE, et al.
The Journal of Biological Chemistry, 280(19), 19410-19418 (2005)
Yan Xing et al.
Scientific reports, 6, 21309-21309 (2016-02-19)
Liver X receptors (LXRs) play essential roles in lipogenesis, anti-inflammatory action and hepatic stellate cells (HSCs) activation in the liver. However, the effects of LXRs on the capillarization of liver sinusoidal endothelial cells (LSECs) in liver fibrosis remain undetermined. Here
Novel putative SREBP and LXR target genes identified by microarray analysis in liver of cholesterol-fed mice
Maxwell KN, et al.
Journal of Lipid Research, 44(11), 2109-2119 (2003)
Inhibition of local macrophage growth ameliorates focal inflammation and suppresses atherosclerosis
Yamada S, et al.
Arteriosclerosis, Thrombosis, and Vascular Biology, 38(5), 994-1006 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service