Skip to Content
Merck

Skip To

SML2283

Lenalidomide

≥98% (HPLC), powder, cereblon binder

Synonym(s):

Lenalidomide, 1-Oxo-4-amino-2-(2,6-dioxopiperidin-3-yl)isoindole, 3-(4-Amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)-2,6-piperidinedione, 3-(4-Amino-1-oxoisoindolin-2-yl)piperidin-2,6-dione, 3-(4-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione, CC 5013, CC-5013

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H13N3O3
CAS Number:
Molecular Weight:
259.26
NACRES:
NA.77
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Lenalidomide, ≥98% (HPLC)

ligand

lenalidomide

assay

≥98% (HPLC)

form

powder

reaction suitability

reagent type: ligand

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1N(C2CCC(NC2=O)=O)CC3=C1C=CC=C3N

InChI

1S/C13H13N3O3/c14-9-3-1-2-7-8(9)6-16(13(7)19)10-4-5-11(17)15-12(10)18/h1-3,10H,4-6,14H2,(H,15,17,18)

InChI key

GOTYRUGSSMKFNF-UHFFFAOYSA-N

Biochem/physiol Actions

Lenalidomide, a derivative of thalidomide, is an immunomodulatory agent that is approved drug for treatment of multiple myeloma. Apparently Lenalidomide is a ligand of ubiquitin E3 ligase cereblon that induces the enzyme to degrade the Ikaros transcription factors IKAROS family zinc finger 1 (IKZF1) and IKZF3. Lenalidomide possess pleiotropic antitumor effects. It is used in the treatment of 5q-deletion associated myelodysplastic syndrome (del(5q)-MDS).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
901558P0018T144
description

≥98% (HPLC)

description

≥95%

description

≥98% (HPLC)

description

≥98%, powder

assay

≥98% (HPLC)

assay

≥95%

assay

≥98% (HPLC)

assay

≥98%

form

powder

form

powder

form

powder

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

solubility

DMSO: 2 mg/mL, clear

solubility

-

solubility

DMSO: ≥14 mg/mL

solubility

DMSO: 20 mg/mL, clear

color

white to beige

color

-

color

yellow

color

white

reaction suitability

reagent type: ligand

reaction suitability

reagent type: ligand

reaction suitability

reagent type: ligand

reaction suitability

reagent type: ligand


Still not finding the right product?

Explore all of our products under Lenalidomide


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Repr. 1B - STOT RE 2

target_organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What are the preparation and storage instructions for Lenalidomide?

    1 answer
    1. In the powder form, this product is stored at 2-8°C. The material is soluble at 2 mg/mL in DMSO. The stability of this product in solution has not been determined. Other sources indicate that aqueous solutions should be prepared fresh daily. This information has not been validated.

      Helpful?

Reviews

No rating value

Active Filters