Skip to Content
Merck
All Photos(4)

Key Documents

N4264

Sigma-Aldrich

4-Nitrophenyl N-acetyl-α-D-galactosaminide

galactosaminide substrate, ≥98% (TLC), powder

Synonym(s):

p-Nitrophenyl 2-acetamido-2-deoxy-α-D-galactopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N2O8
CAS Number:
Molecular Weight:
342.30
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

4-Nitrophenyl N-acetyl-α-D-galactosaminide, substrate for N-acetyl-α-D-galactosaminidase

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

DMF: 25 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H]1[C@@H](O)[C@@H](O)[C@@H](CO)O[C@@H]1Oc2ccc(cc2)[N+]([O-])=O

InChI

1S/C14H18N2O8/c1-7(18)15-11-13(20)12(19)10(6-17)24-14(11)23-9-4-2-8(3-5-9)16(21)22/h2-5,10-14,17,19-20H,6H2,1H3,(H,15,18)/t10-,11-,12+,13-,14+/m1/s1

InChI key

OMRLTNCLYHKQCK-RGDJUOJXSA-N

Biochem/physiol Actions

4-Nitrophenyl N-acetyl-α-D-galactosaminide serves as a chromogenic synthetic substrate for lysosomal enzyme α-N-acetylgalactosaminidase to perform enzyme assays. Upon cleavage the substrate forms a yellow color solution of p-nitrophenol that can be measured at 400nm at pH 10.6.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey B Kaplan et al.
Journal of bacteriology, 185(16), 4693-4698 (2003-08-05)
When cultured in broth, fresh clinical isolates of the gram-negative periodontal pathogen Actinobacillus actinomycetemcomitans form tenaciously adherent biofilm colonies on surfaces such as plastic and glass. These biofilm colonies release adherent cells into the medium, and the released cells can
Simon P J Albracht et al.
BBA clinical, 8, 84-89 (2017-10-25)
This paper presents a new assay to determine the activity of the lysosomal enzyme α-N-acetylgalactosaminidase (Naga, EC 3.2.1.49) in human serum. It is based on the use of a new chromogenic substrate, DNP-α-GalNAc (2,4-dinitrophenyl-N-acetyl-α-D-galactosaminide) and is performed at pH 4.3 and
R D Roer et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 128(4), 683-690 (2001-04-06)
We have previously demonstrated a marked change in sugar moieties of glycoproteins of the cuticle of the blue crab, Callinectes sapidus, between 0.5 and 3 h post-ecdysis. The present study has identified a glycosidase that appears in the cuticle during
S K Chatterjee et al.
Cancer, 49(1), 128-135 (1982-01-01)
Levels and isoenzyme profiles of beta-hexosaminidase were compared in extracts from normal ovarian and ovarian epithelial tumors. The specific activities of beta-hexosaminidase were significantly (P less than 0.001) higher in malignant than in normal ovarian tissues. The enzyme levels of
Jin-Jin Xie et al.
Journal of biomolecular structure & dynamics, 26(4), 509-515 (2008-12-26)
Beta-N-acetyl-D-glucosaminidase (NAGase, EC.3.2.1.52), which catalyzes the cleavage of N-acetylglucosamine polymers, is a composition of chitinase and cooperates with endo-chitinase and exo-chitinase to disintegrate chitin into N-acetylglucosamine (NAG). In this investigation, A NAGase from green crab (Scylla serrata) was purified and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service