Skip to Content
Merck
All Photos(1)

Key Documents

N8759

Sigma-Aldrich

4-Nitrophenyl N-acetyl-α-D-glucosaminide

≥98%, powder

Synonym(s):

p-Nitrophenyl 2-acetamido-2-deoxy-α-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N2O8
CAS Number:
Molecular Weight:
342.30
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

4-Nitrophenyl N-acetyl-α-D-glucosaminide, ≥98%

Quality Level

Assay

≥98%

form

powder

solubility

warm ethanol: acetic acid (1:1): 20 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@@H]1Oc2ccc(cc2)[N+]([O-])=O

InChI

1S/C14H18N2O8/c1-7(18)15-11-13(20)12(19)10(6-17)24-14(11)23-9-4-2-8(3-5-9)16(21)22/h2-5,10-14,17,19-20H,6H2,1H3,(H,15,18)/t10-,11-,12-,13-,14+/m1/s1

InChI key

OMRLTNCLYHKQCK-KSTCHIGDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrophenyl N-acetyl-α-D-glucosaminide is used as a substrate for determining the hydrolysing activities of pnp-glycopyranosides in the gut of larval and adult Psacothea hilaris.

Biochem/physiol Actions

4-Nitrophenyl N-acetyl-α-D-glucosaminide is used as a substrate for analysing β-hexoaminidase activity at 37°C and analysed at 405 nm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diet and carbohydrate digestion in the yellow-spotted longicorn beetle Psacothea hilaris
Scrivener AM, et al.
Journal of Insect Physiology, 43(11), 1039-1052 (1997)
Study of subcellular localization and proteolysis of ataxin-3
Pozzi C, et al.
Neurobiology of Disease, 30(2), 190-200 (2008)
E Frändberg et al.
The Journal of applied bacteriology, 76(3), 259-263 (1994-03-01)
Three methods of quantifying chitinase activity were compared. The activities of crude chitinases of 10 bacterial isolates from different environments were estimated in terms of (1) the release of p-nitrophenol from the chromogenic chito-oligosaccharide analogues, p-nitrophenyl-beta-D-N,N'-diacetylchitobiose, p-nitrophenyl-N-acetyl-beta-D-glucosamine and p-nitrophenyl-beta-D-N,N',N"-triacetylchitotriose, (2)
Ken-ichi Kanno et al.
Lab on a chip, 2(1), 15-18 (2004-04-22)
Glycosidase-promoted hydrolysis was performed in a microreaction channel. The result was compared with the reaction in a micro-test tube. Transgalactosylation on p-nitrophenyl-2-acetamide-2-deoxy-beta-D-glucopyranoside was also examined in a microreaction channel, because transglycosylation is a useful method for oligosaccharide synthesis. We examined
E Chitlaru et al.
The Journal of biological chemistry, 271(52), 33433-33439 (1996-12-27)
The accompanying papers (Keyhani, N. O., and Roseman, S. (1996) J. Biol. Chem. 271, 33414-33424; Keyhani, N. O., and Roseman, S. (1996) J. Biol. Chem. 271, 33425-33432) describe two unique beta-N-acetylglucosaminidases from Vibrio furnissii. A third, ExoII, is reported here.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service