Skip to Content
Merck
All Photos(1)

Key Documents

M264

Sigma-Aldrich

Anti-Glutamate Receptor NMDAR2A (NR2A) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

NR2A Antibody - Anti-Glutamate Receptor NMDAR2A (NR2A) antibody produced in rabbit, Nr2A Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

antigen 180 kDa

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry: 1:1,000-1:2,000
immunoprecipitation (IP): 3 μL using 200 μg of rat brain
western blot: 1:1,000

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRIN2A(2903)
mouse ... Grin2a(14811)
rat ... Grin2a(24409)

General description

Rabbit polyclonal anti-Glutamate Receptor NMDAR2A (NR2A) antibody labels the 180 kDa NR2A subunit of the NMDA receptor. This labeling is blocked by the preadsorption of the antibody with the immunogen. The antibody does not cross-react with the NR2B or NR2C subunits. It reacts with rat, mouse and human tissues.
The N-Methyl-D-Aspartate (NMDA) receptor complex is comprised of 2 types of subunits, NR1 and NR2. Only one type of NR1 subunits has yet been identified while four distinct subunits have been identified for the NR2 receptor, NR2A, NR2B, NR2C, and NR2D. While the NR2 subunits are not functional alone, they combine with the NR1 subunit to produce a variety of different receptor types. A number have studies have also shown that the functional properties of the receptors complexes formed by the NR1 and NR2 subunits are largely determined by the NR2 components of the complex. NMDA receptors are post-synaptic and play important roles in plasticity in the developing and mature central nervous system (CNS). Agonists and antagonists of NMDA receptors have been proposed to be of therapeutic benefit in a number of CNS disorders, including stroke, head injury, epilepsy, pain, and Alzheimer′s disease.

Immunogen

C-terminal portion of NR2A (amino acids 1253-1391).

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Rabbit polyclonal anti-Glutamate Receptor NMDAR2A (NR2A) antibody is used to tag the 180 kDa NR2A subunit of the NMDA receptor for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques such as immunoblotting and immunoprecipitation. It is used as a probe to determine the presence and roles of the 180 kDa NR2A subunit in the assembly and function of N-Methyl-D-Aspartate (NMDA) receptor complexes.

Physical form

Lyophilized from ammonium bicarbonate (5 mM)

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baris Alten et al.
British journal of pharmacology, 175(24), 4450-4463 (2018-09-18)
Children and adolescents are the top consumers of high-fructose corn syrup (HFCS) sweetened beverages. Even though the cardiometabolic consequences of HFCS consumption in adolescents are well known, the neuropsychiatric consequences have yet to be determined. Adolescent rats were fed for
Sighild Lemarchant et al.
International journal of molecular sciences, 23(16) (2022-08-27)
NX210c is a disease-modifying dodecapeptide derived from the subcommissural organ-spondin that is under preclinical and clinical development for the treatment of neurological disorders. Here, using whole-cell patch-clamp recordings, we demonstrate that NX210c increased α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptor (AMPAR)- and GluN2A-containing N-methyl-D-aspartate
L Addis et al.
Scientific reports, 7(1), 66-66 (2017-03-01)
Mutations in the N-methyl-D-aspartate receptor (NMDAR) gene GRIN2A cause epilepsy-aphasia syndrome (EAS), a spectrum of epileptic, cognitive and language disorders. Using bioinformatic and patient data we shortlisted 10 diverse missense mutations for characterisation. We used high-throughput calcium-flux assays and patch
Jennifer Stanic et al.
Nature communications, 6, 10181-10181 (2015-12-19)
NMDA receptor (NMDAR) composition and synaptic retention represent pivotal features in the physiology and pathology of excitatory synapses. Here, we identify Rabphilin 3A (Rph3A) as a new GluN2A subunit-binding partner. Rph3A is known as a synaptic vesicle-associated protein involved in
Luca Franchini et al.
iScience, 19, 927-939 (2019-09-14)
NMDA receptor (NMDAR) subunit composition plays a pivotal role in synaptic plasticity at excitatory synapses. Still, the mechanisms responsible for the synaptic retention of NMDARs following induction of plasticity need to be fully elucidated. Rabphilin3A (Rph3A) is involved in the

Questions

  1. In which volume should I reconstitute the 10 ug antibody?

    1 answer
    1. Anti-Glutamate Receptor NMDAR2A should be reconstituted with 50 ul of sterile phosphate buffered saline (PBS).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service