Skip to Content
Merck
All Photos(4)

Key Documents

G8913

Sigma-Aldrich

Anti-Glutamate Receptor NMDAR1 (NR1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-GluN1, Anti-MRD8, Anti-NDHMSD, Anti-NDHMSR, Anti-NMD-R1, Anti-NMDA1, Anti-NMDAR1, Anti-NR1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 116 kDa

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μL

technique(s)

western blot: 1:2,000 using extract of a rat brain synaptosomal fraction.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRIN1(2902)
mouse ... Grin1(14810)
rat ... Grin1(24408)

General description

N-methyl-D-aspartate receptors (NMDAR1) is mainly expressed in mammalian brain. The gene is located on human chromosome 9q34.3.
NMDAR1 is a subunit of N-methyl D-aspartate (NMDA) receptor which is a subtype of glutamate-gated ion channels with high calcium permeability. NMDA receptors have unique voltage-dependent sensitivity to magnesium and are modified by glycine. It has a pivotal role in neuronal development, pathogenesis of acute and chronic neurodegenerative disorders, synaptic plasticity as well as memory acquisition and learning. Anti-glutamate receptor NMDAR1 (NR1) antibody can be used for studying the expression and function of NMDAR1 in different cell types and tissues as well as to compare its expression pattern with physiological functions or pathological conditions. Rabbit anti-glutamate receptor NMDAR1 (NR1) antibody reacts specifically with NMDAR1 (116kD) derived from rat brain.

Immunogen

synthetic peptide corresponding to the C-terminal region of rat NMDAR1 (amino acids 918-938). This sequence is identical in mouse NR1, human NR1 (short and long forms), and rat NR1 isoforms NR1b, NR1c, and NR1f.

Application

Anti-Glutamate Receptor NMDAR1 (NR1) antibody produced in rabbit has been used in endogenous calmodulin co-immunoprecipitation and immunoprecipitation.
Anti-glutamate receptor NMDAR1 (NR1) antibody can be used as a primary antibody in western blotting. It can also be used to identify NR1 from rat brain synaptosomal fraction by using immunoblotting.

Biochem/physiol Actions

N-methyl-D-aspartate receptor1 (NMDAR1) regulates excitatory synaptic transmission. Abnormal functioning of this receptor is associated with epilepsy and schizophrenia.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diego Peretti et al.
Nature, 518(7538), 236-239 (2015-01-22)
In the healthy adult brain synapses are continuously remodelled through a process of elimination and formation known as structural plasticity. Reduction in synapse number is a consistent early feature of neurodegenerative diseases, suggesting deficient compensatory mechanisms. Although much is known
Ephrin-B2 prevents N-methyl-D-aspartate receptor antibody effects on memory and neuroplasticity
Planaguma J, et al.
Annals of Neurology, 80(3), 388-400 (2016)
Yanil Hepp et al.
Learning & memory (Cold Spring Harbor, N.Y.), 23(8), 427-434 (2016-07-17)
The aim of the present study was to analyze the surface expression of the NMDA-like receptors during the consolidation of contextual learning in the crab Neohelice granulata Memory storage is based on alterations in the strength of synaptic connections between
Single amino acid residue in the M4 domain of GluN1 subunit regulates the surface delivery of NMDA receptors
Kaniakova M, et al.
Journal of Neurochemistry, 123(3), 385-395 (2012)
Jesús Planagumà et al.
Annals of neurology, 80(3), 388-400 (2016-07-12)
To demonstrate that ephrin-B2 (the ligand of EphB2 receptor) antagonizes the pathogenic effects of patients' N-methyl-D-aspartate receptor (NMDAR) antibodies on memory and synaptic plasticity. One hundred twenty-two C57BL/6J mice infused with cerebrospinal fluid (CSF) from patients with anti-NMDAR encephalitis or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service