Skip to Content
Merck
All Photos(2)

Key Documents

M1778

Sigma-Aldrich

Mucin from porcine stomach

Type III, bound sialic acid 0.5-1.5 %, partially purified powder

Synonym(s):

MUC

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61
Pricing and availability is not currently available.

biological source

Porcine stomach

type

Type III

form

partially purified powder

composition

bound sialic acid, 0.5-1.5%

technique(s)

microbiological culture: suitable

solubility

NaOH: soluble 20 mg/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Mucins or mucus glycoproteins are the main macromolecular components of mammalian mucus.[1] It was also used in a study to evaluate the sputum smears concentrated by cyto-centrifugation for detection of acid-fast bacilli.[2]

Application

Mucin from porcine stomach was used in studies on the binding site of the galactose-specific agglutinin PA-IL from Pseudomonas aeruginosa.[3]

Biochem/physiol Actions

Mucus forms the protective cover for all epithelial surfaces. The major structural component of mucus is gel-forming mucins. The mucus layer covering the intestinal epithelium has the MUC2 mucin as its central molecule. MUC2 is produced by goblet cells.[4]

Preparation Note

Prepared according to the method of Glenister, et al., for use in complex growth media for dental plaque bacteria.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenny C Mok et al.
mBio, 11(6) (2020-12-10)
The beneficial human gut bacterium Akkermansia muciniphila provides metabolites to other members of the gut microbiota by breaking down host mucin, but most of its other metabolic functions have not been investigated. A. muciniphila strain MucT is known to use
Anastasia Matthies et al.
Applied and environmental microbiology, 74(15), 4847-4852 (2008-06-10)
The metabolism of isoflavones by gut bacteria plays a key role in the availability and bioactivation of these compounds in the intestine. Daidzein and genistein are the most common dietary soy isoflavones. While daidzein conversion yielding equol has been known
Lihui Feng et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(5), 2622-2633 (2020-01-24)
Human gut microbiota development has been associated with healthy growth but understanding the determinants of community assembly and composition is a formidable challenge. We cultured bacteria from serially collected fecal samples from a healthy infant; 34 sequenced strains containing 103,102
Helen Rose et al.
The Journal of antimicrobial chemotherapy, 63(3), 502-510 (2009-01-21)
The Burkholderia cepacia complex (Bcc) species are important opportunistic pathogens with intrinsic antibiotic resistance. They are also well known as contaminants of disinfectants, yet their biocide susceptibility has not been studied in detail. We investigated Bcc biocide susceptibility and correlated
Leyuan Li et al.
Microbiome, 8(1), 33-33 (2020-03-13)
Human-targeted drugs may exert off-target effects or can be repurposed to modulate the gut microbiota. However, our understanding of such effects is limited due to a lack of rapid and scalable assay to comprehensively assess microbiome responses to drugs. Drugs

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Questions

1–7 of 7 Questions  
  1. Can I reconstitute the mucin so that it forms a saliva/snot-like substance? I can't seem to find any papers that have done this...

    1 answer
    1. Mucins are soluble in water, forming hazy viscous solutions.

      Helpful?

  2. What does the "Type" designation in Mucin from porcine stomach mean?

    1 answer
    1. The type designation does not have any particular biological significance.  It is used to emphasize differences in the listings of this product.

      Helpful?

  3. What are the differences between Mucin from porcine stomach Products M2378 and M1778?

    1 answer
    1. Product No. M2378 is a crude mucin preparation. Product No. M1778 is a partially purified powder prepared according to the method described in Glenister, D.A. and Salmon. K. Microbial Ecol. in Health & Disease 1, 31, (1988). The amount of bound sialic acids are also different. Please refer to the specification sheets on our website.

      Helpful?

  4. Is Product M1778, Mucin from porcine stomach, a purified preparation of mucin?

    1 answer
    1. This product is a partially purified preparation of mucin based on the method of Glenister cited on the product's webpage.

      Helpful?

  5. What is the molecular weight of Product M1778, Mucin from porcine stomach?

    1 answer
    1. This product is partially purified from crude mucin, M2378.  Sigma has not determined the molecular weight of this product. Neither has the sugar and amino acid composition been determined.

      Helpful?

  6. How can I solubilize Product M1778, Mucin from porcine stomach?

    1 answer
    1. Mucin is not readily soluble in aqueous solutions. Turbid "solutions" will form at about 5 mg/mL in 0.1 M acetate buffer, pH 5.0. To try to increase the solubility, one might try further dilution and sonication, but we have not tried this in our labs.

      Helpful?

  7. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service