Skip to Content
Merck
All Photos(1)

Documents

M0880

Sigma-Aldrich

Ethyl methanesulfonate

liquid

Synonym(s):

Ethyl mesylate, Methanesulfonic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO3CH2CH3
CAS Number:
Molecular Weight:
124.16
Beilstein:
773969
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.25

form

liquid

refractive index

n20/D 1.418 (lit.)

bp

85-86 °C/10 mmHg (lit.)

density

1.206 g/mL at 20 °C

SMILES string

CCOS(C)(=O)=O

InChI

1S/C3H8O3S/c1-3-6-7(2,4)5/h3H2,1-2H3

InChI key

PLUBXMRUUVWRLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl methanesulfonate had been used for inducing mutation in mouse embryonic fibroblasts, yeast human embryonic kidney cell lines and human lymphocytes.

Biochem/physiol Actions

Ethyl methanesulfonate is a DNA ethylating agent, mutagenic to plants and animals and carcinogenic in mammals. It has been used as a model alkylating agent in studies of DNA repair processes. EMS induces base substitutions of guanine-cytosine (G/C) to adenine-thymine (A/T). EMS also generates point mutations and single nucleotide polymorphisms in genomes. EMS is potential chemical mutagen used for inducing mutation in rice, wheat and Arabidopsis thaliana.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

212.0 °F

Flash Point(C)

100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Genotoxicity Effect of Ethyl Methanesulfonate on Root Tip Cells of Chilli (Capsicum annuum L.)
Devi AS and Mullainathan L
World Journal of Agricultural Sciences, 7(4), 368-374 (2011)
Novel wine yeast with ARO4 and TYR1 mutations that overproduce `floral?aroma compounds 2-phenylethanol and 2-phenylethyl acetate
Cordente AG, et al.
Applied Microbiology and Biotechnology, 1-12 (2018)
Jinzhu Li et al.
Genes, 11(11) (2020-10-30)
Lateral root development is a complex process regulated by numerous factors. An important role for sugar in lateral root development has been known for a while, but the underlying molecular basis still remains unclear. In this study, we first showed
Kade M Power et al.
PLoS genetics, 16(10), e1009052-e1009052 (2020-10-17)
Ciliary microtubules are subject to post-translational modifications that act as a "Tubulin Code" to regulate motor traffic, binding proteins and stability. In humans, loss of CCP1, a cytosolic carboxypeptidase and tubulin deglutamylating enzyme, causes infantile-onset neurodegeneration. In C. elegans, mutations
Scanning ethyl methanesulphonate effects on the whole genome of Lotus japonicus using second generation sequencing analysis
Mohd-Yusoff NF, et al.
G3 (Bethesda, Md.), g3-114 (2015)

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service