Skip to Content
Merck
All Photos(5)

Key Documents

C9754

Sigma-Aldrich

Colchicine

≥95% (HPLC), powder, antimitotic agent

Synonym(s):

(S)-N-(5,6,7,9-Tetrahydro-1,2,3,10-tetramethoxy-9-oxobenzo[a]heptalen-7-yl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25NO6
CAS Number:
Molecular Weight:
399.44
Beilstein:
2228813
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Colchicine, ≥95% (HPLC), powder

Assay

≥95% (HPLC)

form

powder

color

white to yellow

mp

150-160 °C (dec.) (lit.)

solubility

ethanol: 50 mg/mL

SMILES string

COC1=CC=C2C(=CC1=O)[C@H](CCc3cc(OC)c(OC)c(OC)c23)NC(C)=O

InChI

1S/C22H25NO6/c1-12(24)23-16-8-6-13-10-19(27-3)21(28-4)22(29-5)20(13)14-7-9-18(26-2)17(25)11-15(14)16/h7,9-11,16H,6,8H2,1-5H3,(H,23,24)/t16-/m0/s1

InChI key

IAKHMKGGTNLKSZ-INIZCTEOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Colchicine is a secondary metabolite, which has anti-inflammatory properties. It is used to treat gout, crystal-induced joint inflammation, familial Mediterranean fever, Behçet disease, vasculitides and pericarditis. Colchicine inhibits cell division, intracellular vesicle motility, secretion of cytokines and chemokines. It is implicated in leukocyte diapedesis and lysosomal degranulation. Colchicine also blocks fibroblast proliferation and collagen transport to extracellular space.

Application

Colchicine has been used:
  • in the in vitro sister-chromatid exchange (SCE) assay
  • to study its in vitro antiviral, antibacterial, antifungal and cytotoxic activities
  • in microtubule disruption
  • to analyse its effects on Ca2+ transients from taxol-treated cells

Biochem/physiol Actions

Antimitotic agent that disrupts microtubules by binding to tubulin and preventing its polymerization. Stimulates the intrinsic GTPase activity of tubulin. Induces apoptosis in several normal and tumor cell lines and activates the JNK/SAPK signaling pathway.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage and Stability

This product in powdered format can be stored in room temperature. Colchicine powder will darken upon exposure to light. Colchicine solutions can be sterilized by autoclaving or filtration. Sterilized solutions are stable for at least six months if protected from light. No appreciable colchicine hydrolysis occurred in neutral and slightly alkaline (pH = 8.1) solutions after storage frozen for 2 months.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Muta. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Colchicine-an overview for plant biotechnologists
Ramawat KG and Merillon JM
Bioactive Molecules and Medicinal Plants, 215-232 (2008)
A leukemia fusion protein attenuates the spindle checkpoint and promotes aneuploidy
Boyapati A, et al.
Blood, 109(9), 3963-3971 (2007)
Lynette J Oost et al.
Journal of cachexia, sarcopenia and muscle, 10(3), 630-642 (2019-03-22)
Skeletal muscle is a plastic tissue that adapts to changes in exercise, nutrition, and stress by secreting myokines and myometabolites. These muscle-secreted factors have autocrine, paracrine, and endocrine effects, contributing to whole body homeostasis. Muscle dysfunction in aging sarcopenia, cancer
Genotoxic, cytostatic, antineoplastic and apoptotic effects of newly synthesized antitumour steroidal esters
Karapidaki I, et al.
Mutation Research. Genetic Toxicology and Environmental Mutagenesis, 675(1), 51-59 (2009)
Colchicine today
Niel E and Scherrmann JM
Joint, Bone, Spine : Revue du Rhumatisme, 73(6), 672-678 (2006)

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service