Skip to Content
Merck
All Photos(2)

Documents

72945

Supelco

Ethyl methanesulfonate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Ethyl mesylate, Methanesulfonic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO3CH2CH3
CAS Number:
Molecular Weight:
124.16
Beilstein:
773969
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.418 (lit.)

bp

85-86 °C/10 mmHg (lit.)

density

1.206 g/mL at 20 °C

application(s)

environmental
pharmaceutical

format

neat

storage temp.

−20°C

SMILES string

CCOS(C)(=O)=O

InChI

1S/C3H8O3S/c1-3-6-7(2,4)5/h3H2,1-2H3

InChI key

PLUBXMRUUVWRLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

212.0 °F

Flash Point(C)

100 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interspecific comparison of ethyl methanesulfonate-induced mutation rates in relation to genome size.
A P Schalet
Mutation research, 49(3), 313-340 (1978-03-01)
Elmar Gocke et al.
Toxicology letters, 190(3), 254-265 (2009-10-28)
In order to assess the risk of patients being exposed to an anti-AIDS medication contaminated with EMS we have performed in depth genotoxicity, general toxicity and DMPK investigations. The results of these studies are reported in the accompanying papers of
Nidhi Rawat et al.
BMC plant biology, 12, 205-205 (2012-11-09)
Triticum monococcum L., an A genome diploid einkorn wheat, was the first domesticated crop. As a diploid, it is attractive genetic model for the study of gene structure and function of wheat-specific traits. Diploid wheat is currently not amenable to
Zhenyan Yan et al.
Plant physiology, 150(2), 710-721 (2009-04-28)
GLYCOGEN SYNTHASE KINASE3 (GSK3) is a highly conserved serine/threonine kinase involved in a variety of developmental signaling processes. The Arabidopsis (Arabidopsis thaliana) genome encodes 10 GSK3-like kinases that are clustered into four groups. Forward genetic screens have so far uncovered
Benjamin Hartwig et al.
Plant physiology, 160(2), 591-600 (2012-07-28)
Mapping-by-sequencing (or SHOREmapping) has revitalized the powerful concept of forward genetic screens in plants. However, as in conventional genetic mapping approaches, mapping-by-sequencing requires phenotyping of mapping populations established from crosses between two diverged accessions. In addition to the segregation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service