Skip to Content
Merck
All Photos(1)

Key Documents

L9420

Sigma-Aldrich

Luciferase from Photinus pyralis (firefly)

recombinant, expressed in E. coli, buffered aqueous solution, ≥10×1010 units/mg protein

Synonym(s):

Luciferase firefly

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

Assay

≥98% (SDS-PAGE)

form

buffered aqueous solution

specific activity

≥10×1010 units/mg protein

mol wt

62 kDa

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Firefly luciferase catalyzes the reaction of luciferin with ATP and leads to the production of yellow-green light. The enzyme has a molecular weight of 62kDa and is expressed in peroxisomes. Luciferase is considered as a model to study protein–anesthetic interactions. Firefly luciferase is highly useful in cell biology and molecular biology, as a reporter of gene function and for the quantification of ATP.

Application

Luciferase from Photinus pyralis (firefly) has been used as a component of lysis solution to measure luminescence signals, as part of the study to determine chemical signals which can activate the karrikin insensitive 2 (KAI2) pathway.

Unit Definition

One luciferase enzyme unit will produce one Relative Light Unit (RLU) at 20-25 °C over a 10 second period, measured in 100 μl assay mixture containing 40 pmol ATP and 15 nmol luciferin in Tris-glycine buffer, pH 7.6, using a GloMax 20/20 Luminometer.

Unit Definition Conversion Factor: There are approximately 9000 Relative Light Units (RLU) per one traditional Light Unit that uses a peak height equivalent to 0.02 μCi of 14C in a PPO/POPOP cocktail.

Physical form

Supplied a a solution in 25mM Tris-acetate, pH 7.8 , 1mM EDTA, 1mM DTT, 0.2M ammonium sulfate, 15% glycerol and 30% ethylene glycol

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

231.8 °F

Flash Point(C)

111 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reporter gene-facilitated detection of compounds in Arabidopsis leaf extracts that activate the Karrikin signaling pathway.
Sun Y K, et al.
Frontiers in Plant Science, 7, 1799-1799 (2016)
Crystal structure of firefly luciferase throws light on a superfamily of adenylate-forming enzymes.
Conti E, et al.
Structure, 4(3), 287-298 (1996)
Griffiths A J F, et al.
An Introduction to Genetic Analysis. (2000)
Isabella Nymann Westensee et al.
Small (Weinheim an der Bergstrasse, Germany), 17(24), e2007959-e2007959 (2021-05-11)
Artificial cells (ACs) aim to mimic selected structural and functional features of mammalian cells. In this context, energy generation is an important challenge to be addressed when self-sustained systems are desired. Here, mitochondria isolated from HepG2 cells are employed as
Tanushree Dangi et al.
Cell reports, 42(3), 112167-112167 (2023-03-02)
mRNA vaccines are effective in preventing severe COVID-19, but breakthrough infections, emerging variants, and waning immunity warrant the use of boosters. Although mRNA boosters are being implemented, the extent to which pre-existing immunity influences the efficacy of boosters remains unclear.

Articles

Bioluminescence imaging (BLI) systems allows for high-sensitive and noninvasive monitoring of cell proliferation, activity of signaling pathways and protein-protein interactions in living tissues.

seMpai substrate is suitable for near-infrared BLI in biological experiments, offering high solubility for extended bioluminescent applications.

seMpai substrate is suitable for near-infrared BLI in biological experiments, offering high solubility for extended bioluminescent applications.

seMpai substrate is suitable for near-infrared BLI in biological experiments, offering high solubility for extended bioluminescent applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service