Skip to Content
Merck
All Photos(1)

Key Documents

FLAA

Sigma-Aldrich

Adenosine 5′-triphosphate (ATP) Bioluminescent Assay Kit

for ATP quantitation

Synonym(s):

ATP Bioluminescence Assay, ATP Determination Kit, Luminescent ATP Detection kit

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352202
eCl@ss:
32160414
NACRES:
NA.26

storage temp.

−20°C

General description

The Adenosine 5′-triphosphate (ATP) Bioluminescent Assay Kit may be employed for the quantitative bioluminescent determination of ATP in samples. ATP is consumed and light is emitted when luciferase catalyzes the oxidation of D-luciferin. When ATP is the limiting reagent, the light emitted is proportional to the ATP present in the sample.

Application

Adenosine 5′-triphosphate (ATP) Bioluminescent Assay Kit has also been used in the quantification of ATP in 3D matrixes of human neurons, hepatic cells with ischemia, various bacterial cultures and lysosomes.

Kit Components Only

Product No.
Description

  • FL-AAB Dilution buffer 1 mL/vial

  • FL-AAM Assay mix 1 mL/vial

  • FL-AAS ATP standard 1 mL/vial

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadeshda Andreeva et al.
Applied microbiology and biotechnology, 102(12), 5235-5244 (2018-04-24)
Inorganic polyphosphate (polyP) is a significant regulatory and metabolic compound in yeast cells. We compared polyP content and localization, polyphosphatase activities, and transcriptional profile of polyP-related genes in industrially important methylotrophic yeasts, Hansenula polymorpha and Pichia pastoris. The increased need
F Chen et al.
Journal of clinical microbiology, 32(11), 2791-2800 (1994-11-01)
A bioluminescent assay which employs the luciferin-luciferase ATP-dependent reaction was used to evaluate the viability of populations of Pneumocystis carinii derived from infected rat lungs. Contamination with host cells was reduced by a purification method which involved a combination of
Yuri L Boteon et al.
PloS one, 14(10), e0224066-e0224066 (2019-10-24)
The combination of hypothermic and normothermic machine perfusion (HMP+NMP) of the liver provides individual benefits of both techniques, improving the rescue of marginal organs. The aim of this study was to investigate the effect on the bioenergetic status and the
Evaluation of antimicrobial activities of commercial herb and spice extracts against selected food-borne bacteria
Witkowska A M, et al.
Journal of Food Research, 2(4), 37-37 (2013)
John F C Steele et al.
PloS one, 14(8), e0221226-e0221226 (2019-08-29)
Plant NLRs are modular immune receptors that trigger rapid cell death in response to attempted infection by pathogens. A highly conserved nucleotide-binding domain shared with APAF-1, various R-proteins and CED-4 (NB-ARC domain) is proposed to act as a molecular switch

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service