Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Luciferase from
Photinus pyralis
(firefly)
Synonym(s):
Luciferase firefly
CAS Number:
61970-00-1
Products
Product Category
enzymes (1)
Brand
Sigma-Aldrich (2)
Physical Form
Purity
Recombinant Host
Quality Segment
Application
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
SRE0045
Luciferase from
Photinus pyralis
(firefly),
recombinant, expressed in
E. coli
, lyophilized powder, ≥10×10
10
units/mg protein
Expand
View Pricing
L9420
Luciferase from
Photinus pyralis
(firefly),
recombinant, expressed in
E. coli
, buffered aqueous solution, ≥10×10
10
units/mg protein
Expand
View Pricing