Skip to Content
Merck
All Photos(5)

Key Documents

L5125

Sigma-Aldrich

N-Lauroylsarcosine sodium salt

≥94%

Synonym(s):

N-Dodecanoyl-N-methylglycine sodium salt, Sarkosyl NL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CON(CH3)CH2COONa
CAS Number:
Molecular Weight:
293.38
Beilstein:
5322974
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.32

description

anionic

Assay

≥94%

form

powder

mol wt

micellar avg mol wt 600
average mol wt 600

aggregation number

2

technique(s)

oligo synthesis: suitable

CMC

14.6 mM (20-25°C)

solubility

H2O: 100 mg/mL, clear to slightly hazy, colorless to faintly yellow

SMILES string

[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

InChI

1S/C15H29NO3.Na/c1-3-4-5-6-7-8-9-10-11-12-14(17)16(2)13-15(18)19;/h3-13H2,1-2H3,(H,18,19);/q;+1/p-1

InChI key

KSAVQLQVUXSOCR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

N-Lauroylsarcosine sodium salt has been used as one of the components of the lysing solution in the cornet assay. It has been used for lysing protoplasts in the process of chromosomal DNA isolation form Streptococcus mutans.

Biochem/physiol Actions

N-Lauroylsarcosine is an anionic surfactant which also has protein denaturant potency. In addition, it has been shown as a microbicide for sexually transmitted diseases.

Other Notes

Anionic surfactant.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Houben et al.
Frontiers in molecular neuroscience, 13, 106-106 (2020-08-09)
Alzheimer's disease (AD) is characterized by the accumulation in the brain of intraneuronal aggregates of abnormally and hyperphosphorylated tau proteins and of extracellular deposits of amyloid-β surrounded by dystrophic neurites. Numerous experimental models have shown that tau pathology develops in
N S Andrade et al.
Genetics and molecular research : GMR, 7(4), 1414-1421 (2009-02-19)
Baccharis dracunculifolia De Candole (DC) (Asteraceae) is indigenous throughout southeastern Latin America and is used by local people in traditional medicine. This plant is known to be the source of resin for the highly valued Brazilian green propolis. As no
P W Caufield et al.
Journal of clinical microbiology, 27(2), 274-278 (1989-02-01)
Attempts to study the acquisition, transmission, and other aspects of the natural history of Streptococcus mutans infections in humans have been hampered by limitations and inconsistencies in methods by which phenotypic characteristics of individual isolates are examined. Because most mutans
S Roy et al.
Antimicrobial agents and chemotherapy, 45(6), 1671-1681 (2001-05-17)
The microbicidal efficacies of two anionic surfactants, sodium lauryl sulfate (SLS) and n-lauroylsarcosine (LS), were evaluated in cultured cells and in a murine model of herpes simplex type 2 (HSV-2) intravaginal infection. In vitro studies showed that SLS and LS
Royford Bundi Magiri et al.
Vaccines, 8(2) (2020-05-24)
Vaccination is the most efficient method of protection against influenza infections. However, the rapidly mutating viruses and development of new strains make it necessary to develop new influenza vaccines annually. Hence, vaccines that stimulate cross-protection against multiple influenza subtypes are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service