Skip to Content
Merck
All Photos(1)

Key Documents

61747

Sigma-Aldrich

N-Lauroylsarcosine sodium salt solution

30% aqueous solution, ≥97.0% (HPLC)

Synonym(s):

Sarcosyl, Sarkosyl NL, Sodium N-dodecanoyl-N-methylglycinate, Sodium N-lauroylsarcosinate solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CON(CH3)CH2COONa
CAS Number:
Molecular Weight:
293.38
Beilstein:
5322974
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:
NACRES:
NA.25

description

anionic

Assay

≥97.0% (HPLC)

quality

30% aqueous solution

mol wt

average mol wt 600

aggregation number

2

technique(s)

protein quantification: suitable

pH

7.0-9.0 (25 °C)

CMC

14.6

density

1.033 g/mL at 20 °C

cation traces

Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
K: ≤70 mg/kg
Mg: ≤10 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

InChI

1S/C15H29NO3.Na/c1-3-4-5-6-7-8-9-10-11-12-14(17)16(2)13-15(18)19;/h3-13H2,1-2H3,(H,18,19);/q;+1/p-1

InChI key

KSAVQLQVUXSOCR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

N-Lauroylsarcosine (Sarcosyl) is an ionic surfactant useful in a wide range of solubilization and permeation applications from solubilization of membrane proteins to enhancement of skin permeability in transdermal applications.

Other Notes

Solubilization of membrane proteins; useful for rupturing of eukaryote cells in transcription studies; In the lysis of Clostridium perfringens for rapid extraction of plasmids

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Properties of the polyoma virus transcription complex obtained from mouse nuclei.
R J Shmookler et al.
Virology, 57(1), 122-127 (1974-01-01)
P. Gariglio et al.
Febs Letters, 44, 430-430 (1974)
D E Mahony et al.
Applied and environmental microbiology, 51(3), 521-523 (1986-03-01)
Two rapid methods were evaluated for their extraction of plasmids from Clostridium perfringens. The first method involved lysis of 1 to 2 ml of C. perfringens culture by treatment with hyaluronidase, lysozyme, and sarcosyl. DNA, extracted with phenol-chloroform, was treated
Yeu-Chun Kim et al.
International journal of pharmaceutics, 352(1-2), 129-138 (2007-12-15)
To develop formulations for transdermal drug delivery, this study tested the hypothesis that the anionic surfactant, N-lauroylsarcosine (NLS), and ethanol synergistically increase skin permeability by increasing the fluidity of stratum corneum lipid structure. Skin permeability experiments showed that transdermal delivery
H Wroblewski et al.
Biochimie, 60(4), 389-398 (1978-01-01)
1. Up to 90 per cent of the membrane proteins from Spiroplasma citri could be solubilized with the anionic detergent Sarkosyl (sodium lauroyl-sarcosinate). Maximal solubilization was obtained with 6 to 20 mumoles of of detergent per mg of membrane protein.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service