Skip to Content
Merck
All Photos(3)

Key Documents

L5000

Sigma-Aldrich

N-Lauroylsarcosine, neat

≥95%

Synonym(s):

N-Dodecanoyl-N-methylglycine, Sarkosyl L

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CON(CH3)CH2COOH
CAS Number:
Molecular Weight:
271.40
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

description

anionic

Assay

≥95%

mol wt

average mol wt 600

aggregation number

2

CMC

14.6

SMILES string

CCCCCCCCCCCC(=O)N(C)CC(O)=O

InChI

1S/C15H29NO3/c1-3-4-5-6-7-8-9-10-11-12-14(17)16(2)13-15(18)19/h3-13H2,1-2H3,(H,18,19)

InChI key

BACYUWVYYTXETD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Lauroylsarcosine is an anionic surfactant with an ability to denature proteins. Due to its microbicidal property, N-lauroylsarcosine is being considered as a potent anti-microbicide in topical formulations, especially against sexually transmitted diseases (STDs).

Physical form

May be liquid at room temperature

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manfeng Zhang et al.
PLoS biology, 18(6), e3000748-e3000748 (2020-06-20)
Curli play critical roles in biofilm formation, host cell adhesion, and colonization of inert surfaces in many Enterobacteriaceae. In Escherichia coli, curli biogenesis requires 7 curli-specific gene (csg) products-CsgA through G-working in concert. Of them, CsgG and CsgF are 2
Yoshiaki Furukawa et al.
The Journal of biological chemistry, 286(21), 18664-18672 (2011-04-02)
The transactivation response element (TAR) DNA-binding protein-43 (TDP-43) is a nuclear protein that normally regulates transcription and splicing. Abnormal accumulation of insoluble inclusions containing TDP-43 has been recently reported in the affected tissues of amyotrophic lateral sclerosis (ALS) patients. Here
Kunie Ando et al.
The American journal of pathology, 178(2), 803-816 (2011-02-02)
Many models of human tauopathies have been generated in mice by expression of a human mutant tau with maintained expression of mouse endogenous tau. Because murine tau might interfere with the toxic effects of human mutant tau, we generated a
Victoria C de Leeuw et al.
Reproductive toxicology (Elmsford, N.Y.), 98, 107-116 (2020-09-16)
Human embryonic stem cell neuronal differentiation models provide promising in vitro tools for the prediction of developmental neurotoxicity of chemicals. Such models mimic essential elements of human relevant neuronal development, including the differentiation of a variety of brain cell types
Jian Sun et al.
Nucleic acids research, 43(14), 6827-6846 (2015-07-04)
The transcription factor Pax6 is comprised of the paired domain (PD) and homeodomain (HD). In the developing forebrain, Pax6 is expressed in ventricular zone precursor cells and in specific subpopulations of neurons; absence of Pax6 results in disrupted cell proliferation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service