Skip to Content
Merck
All Photos(1)

Key Documents

L4544

Sigma-Aldrich

Human Laminin

from human fibroblasts, cell culture derived, liquid, 0.5 mg/mL, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75

Product Name

Laminin from human fibroblasts, cell culture derived, liquid, sterile-filtered

biological source

human fibroblasts

Quality Level

sterility

sterile-filtered

form

liquid

packaging

pkg of 100 μL

technique(s)

cell culture | mammalian: suitable

surface coverage

1‑2 μg/cm2

UniProt accession no.

Binding Specificity

Peptide Source: Collagen

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... LAMB1(3912)

Application

Laminin from human fibroblasts has been used:
  • in coating six-well plates for wound healing assay
  • as a substrate in cell adhesion and spreading assay
  • for differentiation of human embryonic stem cells (hESCs) and human induced pluripotent stem cells (hiPSCs) toward dopaminergic neurons
  • in corneal endothelial cell wound healing (migration) assay and corneal endothelial cell barrier assay

Laminin from human fibroblasts is recommended for use as a cell culture substratum at 1-2 μg/cm2. The optimal concentration does depend on cell type as well as the application or research objectives.

Biochem/physiol Actions

Laminin proteins are integral components of structural scaffolding in animal tissues. They associate with type IV collagen via entactin and perlecan and bind to cell membranes through integrin receptors, dystroglycan glycoprotein complexes and Lutheran blood group glycoproteins. Laminin has active domains for collagen binding, cell adhesion, heparin binding, and neurite outgrowth fragment. Laminin supports growth and differentiation of many cell types including epithelial, endothelial, neural, muscle and liver cells.

Components

Laminin is an extracellular matrix multidomain trimeric glycoprotein, and is the main non-collagenous component of basal lamina that supports adhesion, proliferation and differentiation. Laminin is composed of both A, B1 and B2 chains, which are connected by many disulfide bonds. This laminin product is produced by human fibroblasts and epithelial cells in a co-culture system and then purified biochemically.

Caution

It is recommended to store this product at -70°C, where it will remain stable for two years.

Preparation Note

This product is supplied at a concentration of 0.5 mg/mL in TBS. Thaw this solution slowly before use at 2-8°C to avoid gel formation. If at any time prior to that, the laminin is allowed to warm up prematurely, gelling can occur. Moreover, even allowing it to be stored at -20C can induce gelling or significantly reduce its activity or both." For use as a coating, dilute in a HBSS, coat culture surface with a minimal volume and incubate at 37°C for 1-2 hours. Wash 3 times with HBBS before plating cells. Laminin coatings can be stored for one month at 2-8°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient neural conversion of human pluripotent stem cells in adherent and animal-free conditions
Lukovic D, et al.
Stem Cells Translational Medicine, 6(4), 1217-1226 (2017)
Elucidating the molecular basis of PPCD: Effects of decreased ZEB1 expression on corneal endothelial cell function
Zakharevich M, et al.
Molecular Vision, 740-740 (2017)
Serum-induced differentiation of glioblastoma neurospheres leads to enhanced migration/invasion capacity that is associated with increased MMP9
Joseph JV, et al
PLoS ONE, 10(12), e0145393-e0145393 (2015)
The depletion of MARVELD1 leads to murine placenta accreta via integrin I?4-dependent trophoblast cell invasion.
Chen, et al.
Journal of Cellular Physiology, 233, 2257-2269 (2017)
Danielle K Lewis et al.
Aging cell, 7(6), 836-849 (2008-09-10)
Astrocytes comprise a large proportion of the central nervous system support cells and play a critical role in neural injury and repair. The present study examined the impact of ovarian aging using an ex vivo model system, where astrocytes were

Articles

The extracellular matrix (ECM) and its attachment factor components are discussed in this article in relation to their function in structural biology and their availability for in vitro applications.

The extracellular matrix (ECM) and its attachment factor components are discussed in this article in relation to their function in structural biology and their availability for in vitro applications.

The extracellular matrix (ECM) and its attachment factor components are discussed in this article in relation to their function in structural biology and their availability for in vitro applications.

The extracellular matrix (ECM) and its attachment factor components are discussed in this article in relation to their function in structural biology and their availability for in vitro applications.

See All

Protocols

Coating surfaces with laminin for culturing cells requires specific conditions for optimal results. Protocols for coating coverslips to culture neurospheres and general cell culture are included.

Coating surfaces with laminin for culturing cells requires specific conditions for optimal results. Protocols for coating coverslips to culture neurospheres and general cell culture are included.

Coating surfaces with laminin for culturing cells requires specific conditions for optimal results. Protocols for coating coverslips to culture neurospheres and general cell culture are included.

Coating surfaces with laminin for culturing cells requires specific conditions for optimal results. Protocols for coating coverslips to culture neurospheres and general cell culture are included.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service