Skip to Content
Merck
All Photos(7)

Key Documents

L9393

Sigma-Aldrich

Anti-Laminin (LAMA1) antibody

enhanced validation

rabbit polyclonal

Synonym(s):

Laminin Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

animal, human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

concentration

0.5 mg/mL

technique(s)

dot blot: 1:1,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:30 using human and animal tissues
microarray: suitable

UniProt accession no.

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAMA1(284217)
mouse ... Lama1(16772)

General description

Laminin is a ubiquitous non-collagenous connective tissue glycoprotein that is a major constituent of basement membranes. Laminins are extracellular matrix proteins. The LAMA1 (laminin subunit α 1) gene is mapped to human chromosome 18p11. The antibody is isolated from antiserum by immunospecific methods of purification. Antigen specific isolation removes essentially all rabbit serum proteins, including immunoglobulins that do not specifically bind to laminin.

Specificity

Specificity of the anti-laminin antibodies is determined by indirect immunofluorescent labeling of formalin-fixed, paraffin-embedded human or animal tissue sections, and by dot blot immunoassay. By indirect immunofluorescence the antibody demonstrates specific basement membrane staining of enzymatically unmasked human and animal tissue. In the dot blot immunoassay the rabbit anti-laminin antibody reacts with laminin but not with fibronectin, vitronectin, collagen IV, or chondroitin sulfate types A, B, and C. The affinity isolated antibody to laminin will react with laminin of human, mammal, avian, reptilian, and amphibian sources.

Immunogen

Laminin isolated from the basement membrane of Englebreth Holm-Swarm (EHS) mouse sarcoma.

Application

Anti-Laminin antibody has been used in immunohistochemical staining and immunohistochemistry.
Rabbit Anti-Laminin antibody may be used in immunohistochemistry for marking blood vessel walls in different species, classification of various disease processes involving basement membranes, identification of the origin of human tumors and their classification, and for distinguishing between non-invasive and invasive lesions. The antibody may be used to monitor levels of laminin in biological fluids and for experimental production of basement membrane lesions in vivo.
A working dilution of at least 1:1,000 was determined by a dot blot immunoassay using laminin at 50 ng per dot.
A working dilution of at least 1:25 was determined by indirect immunohistology using formalin-fixed, paraffin-embedded human and animal tissues.

Biochem/physiol Actions

LAMA1 (laminin subunit α 1) is involved in several diseases like cancer, infections, inflammatory diseases and autoimmune disorders. Mutations in LAMA1 result in cerebellar dysplasia and cysts with and without retinal dystrophy. LAMA1 is also essential in the assembly of basement membrane and early growth of embryo.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zui Zhang et al.
Nature communications, 10(1), 3561-3561 (2019-08-10)
Protein corona presents a major obstacle to bench-to-bedside translation of targeted drug delivery systems, severely affecting targeting yields and directing unfavorable biodistribution. Corona-mediated targeting provides a new impetus for specific drug delivery by precisely manipulating interaction modes of functional plasma
Solving an enigma: Arterial pole development in the zebrafish heart
Adrian CG, et al.
Developmental Biology, 290(2), 265-276 (2006)
Prostatic Intraepithelial Neoplasia in Genetically Engineered Mice
Jae-Hak P, et al.
The American Journal of Pathology, 161(2), 727-735 (2002)
Autoimmunity against laminins.
Florea F, et al.
Clinical Immunology (Orlando, Fla.), 39-52 (2016)
Mutations in LAMA1 Cause Cerebellar Dysplasia and Cysts with and without Retinal Dystrophy
Kimberly AA, et al.
American Journal of Human Genetics, 95(2), 227-234 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service