Skip to Content
Merck
All Photos(1)

Key Documents

I0140

Sigma-Aldrich

Anti-Human IgM (μ-chain specific) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

indirect ELISA: 1:100,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

IgM is a class of antibodies containing a pentamer of five IgG equivalents with 10 Fab fragments, joined by a single cross-linking peptide (J-chain). It constitutes 15% of the normal adult immunoglobulins.

Specificity

The antibody preparation is specific for human IgM when tested against purified human IgA, IgG, IgM, Bence Jones kappa, and lambda myeloma proteins.

Application

Anti-Human IgM (μ-chain specific) antibody produced in rabbit was used as secondary antibody in immunoblotting and indirect ELISA.

Biochem/physiol Actions

IgM antibodies are present as pentamers in the serum and are produced in response to antigens.
Immunoglobulin M (IgM) antibodies appear early in the course of infections. IgM is an initial immunoglobulin produced in the developing fetus as a result of the immune response during the second trimester of pregnancy. It is then substituted by IgG. Therefore, the diagnosis of specific antibodies of the IgM class is useful in detecting chronic infections. IgM is essential for the fetal and neonatal host defense system. IgM cannot cross the placenta. Hence, the presence of IgM might indicate an intrauterine viral infection. IgM antibodies are responsible for the agglutination of red blood cells in mismatched blood transfusions. The level of IgM may vary with the status of disease or infection.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as preservative

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots at –20 °C. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immunology of the Fetus and Newborn
Avery's Diseases of the Newborn, 445-467 null
Overview of the Immune Response
The Journal of Allergy and Clinical Immunology null
J Raper et al.
Infection and immunity, 67(4), 1910-1916 (1999-03-20)
Natural resistance of humans to the cattle pathogen Trypanosoma brucei brucei has been attributed to the presence in human serum of nonimmune factors that lyse the parasite. Normal human serum contains two trypanosome lytic factors (TLFs). TLF1 is a 500-kDa
E J Wiersma et al.
Journal of immunology (Baltimore, Md. : 1950), 160(12), 5979-5989 (1998-06-24)
Previous studies have discerned two forms of polymeric mouse IgM: moderately cytolytic (complement-activating) pentamer, which contains J chain, and highly cytolytic hexamer, which lacks J chain. To investigate the relationships among polymeric structure, J chain content, and cytolytic activity, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service