Skip to Content
Merck
All Photos(1)

Documents

I2386

Sigma-Aldrich

Anti-Human IgM (μ-chain specific) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-IgM produced in Goat, Goat Anti-Human IgM, IgM Detection Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

indirect ELISA: 1:20,000-1:40,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgM antibodies are present as pentamers in the serum and are produced in response to antigens.

Application

Anti-Human IgM (μ-chain specific) antibody produced in goat has been used in enzyme linked immunosorbent assay (ELISA).

Biochem/physiol Actions

Immunoglobulin M (IgM) antibodies are the primary ones to be produced in response to antigen. They bind with low affinity but exhibit a broad specificity to a variety of antigens. IgM constitutes 15% of adult immunoglobulins. IgM is important for fetal and neonatal host defenses for several reasons. IgM molecule efficiently activates classical pathway of complement. Its pentameric structure helps in accomodating multivalent ligand binding. IgM has a prominent role in clearance of invading microorganisms from serum.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anti-protein immunoglobulin M responses to pneumococcus are not associated with aging
German EL, et al.
Pneumonia (Nathan Qld.), 10(1), 5-5 (2018)
J Ollila et al.
Genes and immunity, 4(5), 343-350 (2003-07-09)
The development of adaptive immunity and responses to foreign molecules and organisms relies on the highly regulated production of hundreds of proteins. B-cell maturation, from committed progenitors to terminally differentiated plasma cells, is a multistep process that requires the ordered
Yang Li et al.
Cell reports, 34(13), 108915-108915 (2021-03-25)
To fully decipher the immunogenicity of the severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) Spike protein, it is essential to assess which part is highly immunogenic in a systematic way. We generate a linear epitope landscape of the Spike protein
Fredy Brice Nemg Simo et al.
Intervirology, 61(6), 265-271 (2019-05-03)
On May 2017, a case of dengue serotype 1 was detected and confirmed through routine surveillance in a traveler returning from Kribi, a seaside town of Southern Cameroon. This study aimed at confirming the circulation of dengue virus (DENV) in
Johannes U Ammann et al.
Current protocols in protein science, 75, 19-19 (2014-02-11)
Discovery of binding partners for immunoglobulin molecules expressed by cells of the immune system is an important topic of current research. However, many ligand-receptor interactions are of low affinity, and hence detection is refractory to most established protocols. We evaluated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service