Skip to Content
Merck
All Photos(1)

Documents

HPA039936

Sigma-Aldrich

Anti-MAP3K12 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Dlk, Anti-Mekk12, Anti-Mitogen-activated protein kinase kinase kinase 12, Anti-Muk, Anti-Zpk, Anti-Zpkp1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

GAKGEPPPPVGPGEGVGLLGTGREGTSGRGGSRAGSQHLTPAALLYRAAVTRSQKRGISSEEEEGEVDSEVELTSSQRWPQSLNMRQSLSTFSSENPSDGEEGTASEPSPSGTPEVGSTNTDERPDERSDDMCSQGSEIPLDP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAP3K12(7786)

General description

MAP3K12 (mitogen-activated protein kinase kinase kinase 12) belongs to the serine/threonine protein kinase family. It is also known as DLK (dual leucine zipper kinase). MAP3K12 is located on human chromosome 12q13.

Immunogen

mitogen-activated protein kinase kinase kinase 12 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MAP3K12 (mitogen-activated protein kinase kinase kinase 12) participates in development, cell differentiation, apoptosis and neuronal response to injury. DLK (dual leucine zipper kinase) serves as an upstream activator of the mitogen-activated protein kinase (MAPK) pathways. It may be essential for proliferation of cells. MAP3K12 is involved in the growth of neurons and helps in axon regeneration. It also participates in the pathogenesis of neurodegenerative disease and diabetes mellitus.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71425

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of the 12q amplicons in lipomatous soft tissue tumors by multiplex ligation-dependent probe amplification-based copy number analysis.
Creytens D, et al.
Anticancer Research, 35(4), 1835-1842 (2015)
Dual leucine zipper kinase (MAP3K12) modulators: a patent review (2010?2015).
Oetjen E and Lemcke T
Expert Opinion on Therapeutic Patents, 26(5), 607-616 (2016)
Loss of DLK expression in WI-38 human diploid fibroblasts induces a senescent-like proliferation arrest.
Daviau A, et al.
Biochemical and Biophysical Research Communications, 413(2), 282-287 (2011)
Jingwen Niu et al.
Cell reports, 33(7), 108365-108365 (2020-11-19)
After optic nerve crush (ONC), the cell bodies and distal axons of most retinal ganglion cells (RGCs) degenerate. RGC somal and distal axon degenerations were previously thought to be controlled by two parallel pathways, involving activation of the kinase dual
Dale D O Martin et al.
Scientific reports, 9(1), 3632-3632 (2019-03-08)
After axonal insult and injury, Dual leucine-zipper kinase (DLK) conveys retrograde pro-degenerative signals to neuronal cell bodies via its downstream target c-Jun N-terminal kinase (JNK). We recently reported that such signals critically require modification of DLK by the fatty acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service