Skip to Content
Merck
All Photos(1)

Documents

H5653

Sigma-Aldrich

8-Hydroxy-2′-deoxyguanosine

≥98% (TLC), DNA damage biomaker

Synonym(s):

8-Oxo-7,8-dihydro-2′-deoxyguanosine, 8-Oxo-dG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O5
CAS Number:
Molecular Weight:
283.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

8-Hydroxy-2′-deoxyguanosine, ≥98% (TLC)

Quality Level

Assay

≥98% (TLC)

storage temp.

2-8°C

SMILES string

O=C1C(NC(N2C3CC(O)C(CO)O3)=O)=C2NC(N)=N1

InChI

1S/C10H13N5O5/c11-9-13-7-6(8(18)14-9)12-10(19)15(7)5-1-3(17)4(2-16)20-5/h3-5,16-17H,1-2H2,(H,12,19)(H3,11,13,14,18)

InChI key

HCAJQHYUCKICQH-UHFFFAOYSA-N

Application

8-Hydroxy-2′-deoxyguanosine has been used:
  • as a standard to analyse the oxidative DNA damage in the placenta of normal and abnormal pregnancies by electrochemical detection
  • as a standard for high performance liquid chromatography (HPLC) with electrochemical detection (ECD) measurement
  • as a standard for HPLC and as a urinary marker of oxidative stress in order to assess the potential antioxidant activity of green coffee bean extract (GCBE) in vivo
  • as a standard for LC/MS/MS analysis

Biochem/physiol Actions

A marker compound typically indicative of DNA damage associated with mutagenesis and carcinogenesis
The levels of 8-hydroxy-2′-deoxyguanosine helps in evaluating oxidative stress and DNA damage in individuals suffering from laryngeal cancer. Hence it serves as a major biomarker of DNA damage, induced by reactive oxygen species (ROS).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisabeth Müllner et al.
Molecular nutrition & food research, 57(2), 328-338 (2012-11-14)
Type 2 diabetes is a multifactorial disease associated with increased oxidative stress, which may lead to increased DNA damage. The aim of this study was to investigate the effect of a healthy diet on DNA oxidation in diabetics and nondiabetics.
Rachel Yong et al.
PloS one, 8(8), e73428-e73428 (2013-08-31)
NADPH oxidase 4 (Nox4) is reported to be the major source of reactive oxygen species (ROS) in the kidneys during the early stages of diabetic nephropathy. It has been shown to mediate TGFβ1-induced differentiation of cardiac fibroblasts into myofibroblasts. Despite
Pan Chen et al.
Life sciences, 232, 116594-116594 (2019-06-25)
Sleep deficiency has been reported to be associated with some oral health problems. Oral ulcers are very common lesions of the oral mucosa, which severely impact patients' quality of life. However, the association between sleep deficiency and the oral ulcer
Measuring oxidative DNA damage with 8-hydroxy-2?-deoxyguanosine levels in patients with laryngeal cancer
Mazlumoglu M R, et al.
The Annals of Otology, Rhinology, and Laryngology, 126(2), 103-109 (2017)
Liang Cui et al.
Chemical research in toxicology, 26(2), 195-202 (2012-11-13)
Oxidative damage to DNA has many origins, including irradiation, inflammation, and oxidative stress, but the chemistries are not the same. The most oxidizable base in DNA is 2-deoxyguanosine (dG), and the primary oxidation products are 8-oxodG and 2-amino-imidazolone. The latter

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service