Skip to Content
Merck
All Photos(6)

Documents

D7400

Sigma-Aldrich

2′-Deoxyadenosine monohydrate

≥99%

Synonym(s):

9-(2-Deoxy-β-D-ribofuranosyl)adenine, Adenine deoxyriboside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O3 · H2O
CAS Number:
Molecular Weight:
269.26
Beilstein:
91015
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

≥99%

form

powder

solubility

water: 25 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

[H]O[H].Nc1ncnc2n(cnc12)[C@H]3C[C@H](O)[C@@H](CO)O3

InChI

1S/C10H13N5O3/c11-9-8-10(13-3-12-9)15(4-14-8)7-1-5(17)6(2-16)18-7/h3-7,16-17H,1-2H2,(H2,11,12,13)/t5-,6+,7+/m0/s1

InChI key

OLXZPDWKRNYJJZ-RRKCRQDMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2′-Deoxyadenosine monohydrate has been used:
  • as a component for nucleoside supplementation
  • as a standard for to estimate DNA global methylation rate in proliferating oil palm embryogenic suspensions
  • to reverse sterile alpha motif (SAM) and histidine-aspartate (HD) domain-containing protein (SAMHD1)
  • inhibition of human immunodeficiency virus (HIV-1) and hepatitis B virus (HBV)

Biochem/physiol Actions

2′-Deoxyadenosine (2′-dAdo), a deoxyribonucleoside, may be used by some cells as an energy source under energy stress conditions and to affect cAMP levels. 2′-dAdo is used in comparison studies of the functions of adenosine analogues on various biological processes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Magnus et al.
International journal of fertility, 38(2), 123-128 (1993-03-01)
To find differences in effect on sperm motility of agents that increase intracellular cAMP: manganese ion, methyl-isobutyl-xanthine (MIX), 2-deoxyadenosine, glucose, and Mn-MIX and Mn-glucose. Nine men with asthenozoospermia vs. fertile donors. Sperm was washed in Hepes-buffered saline, motility tested by
M Nakamura et al.
The Journal of biological chemistry, 257(23), 13945-13950 (1982-12-10)
Regulation of glucose metabolism in glycolysis by round spermatids was studied. Assay of activities of 11 glycolytic enzymes in cell-free spermatid extracts showed that hexokinase, phosphofructokinase, and glyceraldehyde-3-phosphate dehydrogenase had the lowest activities. When the cells were incubated with glucose
Nucleoside supplementation modulates mitochondrial DNA copy number in the dguok-/- zebrafish
Munro B, et al.
Human Molecular Genetics, 28(5), 796-803 (2018)
Variations in genomic DNA methylation during the long-term in vitro proliferation of oil palm embryogenic suspension cultures
Rival A, et al.
Plant Cell Reports, 32(3), 359-368 (2013)
Don E Farthing et al.
Scientific reports, 7(1), 4043-4043 (2017-06-24)
Deuterated water (

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service