Skip to Content
Merck
All Photos(5)

Key Documents

D7145

Sigma-Aldrich

2′-Deoxyguanosine monohydrate

99-100%

Synonym(s):

2′-Deoxyguanosine hydrate, 9-(2-Deoxy-β-D-ribofuranosyl)guanine, Guanine-2′-deoxyriboside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O4 · H2O
CAS Number:
Molecular Weight:
285.26
Beilstein:
39814
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

99-100%

form

powder

solubility

1 M NH4OH: 50 mg/mL, clear, colorless

SMILES string

O.NC1=Nc2c(ncn2[C@H]3C[C@H](O)[C@@H](CO)O3)C(=O)N1

InChI

1S/C10H13N5O4.H2O/c11-10-13-8-7(9(18)14-10)12-3-15(8)6-1-4(17)5(2-16)19-6;/h3-6,16-17H,1-2H2,(H3,11,13,14,18);1H2/t4-,5+,6+;/m0./s1

InChI key

LZSCQUCOIRGCEJ-FPKZOZHISA-N

Looking for similar products? Visit Product Comparison Guide

Application

2′-Deoxyguanosine monohydrate has been used as a nucleoside supplement:
  • to study its effect on mitochondrial DNA copy number in deoxyguanosine kinase (dguok) mutant zebrafish,
  • in tissue culture medium for deoxyribonucleotide triphosphate (dNTP) synthesis,
  • in RPMI (Roswell Park Memorial Institute)-1640 medium to eliminate the endogenous thymocytes

Biochem/physiol Actions

Deoxyguanosine (dG) is a purine nucleoside that upon sequential phosphorylation (kinases) forms deoxyguanosine triphosphate (dGTP) which is used by DNA polymerases and reverse transcriptases to synthesize DNA(s). Deoxyguanosine is the most electron-rich of the four canonical bases and includes many nucleophilic sites which are susceptible to oxidative damage. This makes deoxyguanosine and its oxidized derivatives useful reagents to study mechanisms of oxidative damage to nucleosides and nucleotides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zimu Deng et al.
Methods in molecular biology (Clifton, N.J.), 1323, 151-158 (2015-08-22)
Reconstituted thymus organ culture is based on fetal thymus organ culture (FTOC). Purified thymocyte populations, from genetically modified mice or even from other species, are cultured in vitro with thymic lobes depleted of their endogenous thymocytes (by 2'-deoxyguanosine treatment) to
Kasper Broedbaek et al.
Free radical biology & medicine, 51(8), 1473-1479 (2011-08-09)
The increasing prevalence of diabetes together with the associated morbidity and mortality calls for additional preventive and therapeutic strategies. New biomarkers that can be used in therapy control and risk stratification as alternatives to current methods are needed and can
Takeji Takamura-Enya et al.
Bioorganic & medicinal chemistry letters, 21(14), 4206-4209 (2011-06-21)
BODIPY-modified 2'-deoxyguanosine was synthesized for use as a detection reagent for genotoxic compounds. BODIPY-FL is a well known fluorescence reagent whose fluorescent light emission diminishes near a guanine base by a photo-induced electron transfer process. We attached BODIPY-Fl to the
Benjamin Munro et al.
Human molecular genetics, 28(5), 796-803 (2018-11-15)
Deoxyguanosine kinase (dGK) is an essential rate-limiting component of the mitochondrial purine nucleotide salvage pathway, encoded by the nuclear gene encoding deoxyguanosine kinase (DGUOK). Mutations in DGUOK lead to mitochondrial DNA (mtDNA) depletion typically in the liver and brain, causing
José Pedro F Angeli et al.
Free radical biology & medicine, 51(2), 503-515 (2011-05-24)
Epidemiological studies have indicated that Western diets are related to an increase in a series of malignancies. Among the compounds that are credited for this toxic effect are heme and lipid peroxides. We evaluated the effects of hemoglobin (Hb) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service