Skip to Content
Merck
All Photos(1)

Key Documents

281042

Sigma-Aldrich

Tetradecafluorohexane

99%

Synonym(s):

Perfluorohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)4CF3
CAS Number:
Molecular Weight:
338.04
Beilstein:
1802113
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.252 (lit.)

bp

58-60 °C (lit.)

mp

-74 °C (lit.)

density

1.669 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C6F14/c7-1(8,3(11,12)5(15,16)17)2(9,10)4(13,14)6(18,19)20

InChI key

ZJIJAJXFLBMLCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetradecafluorohexane in the gas phase reacts spontaneously with lithium amalgam, to give a solid and intimate mixture of lithium fluoride and elemental polymeric carbon with a small amount of superstoichiometric lithium.

Application

Tetradecafluorohexane has been used:
  • as a fluorocarbon organic solvent in the preparation of temperature-induced phase-separation solution
  • to investigate boiling heat transfer mechanisms
  • as a photosensitizer in fluorous biphasic singlet oxygenation
  • as a novel reaction medium for photooxidation reactions

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic Communications, 26, 1861-1861 (1996)
Journal of the American Chemical Society, 118, 5312-5312 (1996)
Journal of Heat Transfer, 118, 429-429 (1996)
Michael P Grubb et al.
Nature chemistry, 8(11), 1042-1046 (2016-10-22)
Spectroscopically observing the translational and rotational motion of solute molecules in liquid solutions is typically impeded by their interactions with the solvent, which conceal spectral detail through linewidth broadening. Here we show that unique insights into solute dynamics can be
Koichi Kitaguchi et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 30(6), 687-690 (2014-06-13)
A fluorocarbon and hydrocarbon organic solvent mixture is known as a temperature-induced phase-separation solution. When a mixed solution of tetradecafluorohexane as a fluorocarbon organic solvent and hexane as a hydrocarbon organic solvent (e.g., 71:29 volume ratio) was delivered in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service