Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Heptacosafluorotributylamine
Synonym(s):
PFTBA, Perfluorotributylamine, Tris(nonafluorobutyl)amine
Linear Formula:
N(CF
2
CF
2
CF
2
CF
3
)
3
CAS Number:
311-89-7
Molecular Weight:
671.09
EC Number:
206-223-1
Products
Brand
Sigma-Aldrich (1)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Format
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
H5262
Heptacosafluorotributylamine,
liquid
Expand
View Pricing
77299
Heptacosafluorotributylamine,
analytical standard, suitable for mass spectrometry (MS)
Expand
View Pricing