Skip to Content
Merck
All Photos(6)

Documents

G7000

Sigma-Aldrich

α-D-Glucose 1-phosphate disodium salt hydrate

≥97% (Enzymatic Purity, anhydrous)

Synonym(s):

α-D-Glucopyranose 1-phosphate, Cori ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11O9PNa2 · xH2O
CAS Number:
Molecular Weight:
304.10 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Assay

≥97% (Enzymatic Purity, anhydrous)

form

powder

impurities

≤0.1 mol % glucose
≤0.5 mol % α-D-glucose 1,6-diphosphate

color

white

solubility

water: 50 mg/mL, clear, colorless

cation traces

Na: 13.5-16.7% (anhydrous)

application(s)

agriculture

storage temp.

−20°C

SMILES string

[Na+].[Na+].[H]O[H].OC[C@H]1O[C@H](OP([O-])([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H13O9P.2Na.H2O/c7-1-2-3(8)4(9)5(10)6(14-2)15-16(11,12)13;;;/h2-10H,1H2,(H2,11,12,13);;;1H2/q;2*+1;/p-2/t2-,3-,4+,5-,6-;;;/m1.../s1

InChI key

YWAUQXHOCBBYLP-FBNUBEQJSA-L

Looking for similar products? Visit Product Comparison Guide

General description

α-D-Glucose 1-phosphate is converted into D-glucose-6-phosphate by the enzyme phosphoglucomutase.

Biochem/physiol Actions

Glucose-1-phosphate (G1P) is produced from glycogen during glycogenolysis by the actions of glycogen phosphorylase. Conversion to glucose-6-phosphate (G6P) by phosphoglucomutase allows for entry of the glucose molecule into metabolic pathways such as glycolysis. During glycogenesis, G6P is converted to G1P by the actions of phosphoglucose isomerase.

Preparation Note

Prepared by a modification of the procedure of McCready, R.M., et al., J. Am. Chem. Soc., 66, 560 (1944).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Yang et al.
The Biochemical journal, 429(3), 533-543 (2010-05-21)
The diverse types of glycoconjugates synthesized by trypanosomatid parasites are unique compared with the host cells. These glycans are required for the parasite survival, invasion or evasion of the host immune system. Synthesis of those glycoconjugates requires a constant supply
Jef Van der Borght et al.
Biotechnology journal, 5(9), 986-993 (2010-08-28)
β-D-Glucose-1-phosphate (βGlc1P) is an efficient glucosyl donor for both enzymatic and chemical glycosylation reactions but is currently very costly and not available in large amounts. This article provides an efficient production method of βGlc1P from trehalose and phosphate using the
Kyra-Melinda Alexacou et al.
Bioorganic & medicinal chemistry, 18(22), 7911-7922 (2010-10-16)
Glycogen phosphorylase (GP) is a promising target for the treatment of type 2 diabetes. In the process of structure based drug design for GP, a group of 15 aromatic aldehyde 4-(β-d-glucopyranosyl)thiosemicarbazones have been synthesized and evaluated as inhibitors of rabbit
Joerg Fettke et al.
Plant physiology, 155(4), 1723-1734 (2010-12-01)
Almost all glucosyl transfer reactions rely on glucose-1-phosphate (Glc-1-P) that either immediately acts as glucosyl donor or as substrate for the synthesis of the more widely used Glc dinucleotides, ADPglucose or UDPglucose. In this communication, we have analyzed two Glc-1-P-related
Daniel Decker et al.
Phytochemistry, 79, 39-45 (2012-05-04)
UDP-Glc pyrophosphorylase (UGPase) is an essential enzyme responsible for production of UDP-Glc, which is used in hundreds of glycosylation reactions involving addition of Glc to a variety of compounds. In this study, barley UGPase was characterized with respect to effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service