Skip to Content
Merck
All Photos(5)

Key Documents

G6875

Sigma-Aldrich

α-D-Glucose 1-phosphate dipotassium salt hydrate

≥97% (HPLC)

Synonym(s):

α-D-Glucopyranose 1-phosphate, Cori ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11K2O9P · xH2O
CAS Number:
Molecular Weight:
336.32 (anhydrous basis)
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

synthetic

Assay

≥97% (HPLC)

form

powder

impurities

glucose, essentially free

color

white

solubility

water: slightly soluble 50 g/L

storage temp.

−20°C

SMILES string

[K+].[K+].[H]O[H].OC[C@H]1O[C@H](OP([O-])([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H13O9P.2K.H2O/c7-1-2-3(8)4(9)5(10)6(14-2)15-16(11,12)13;;;/h2-10H,1H2,(H2,11,12,13);;;1H2/q;2*+1;/p-2/t2-,3-,4+,5-,6-;;;/m1.../s1

InChI key

VOQGDSVKCMGEFO-FBNUBEQJSA-L

Looking for similar products? Visit Product Comparison Guide

General description

α-D-Glucose 1-phosphate is the α-anomeric form of glucose which contains a phosphate group on the primary carbon. It can be converted into the deoxysugar CDP-glucose by the enzyme α-D-Glucose-1-phosphate cytidylyltransferase.

Linkage

Formerly listed as Grade I.

Preparation Note

Prepared by a modification of the procedure of McCready, R.M., et al., J. Am. Chem. Soc., 66, 560 (1944).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bijay Singh et al.
Protein engineering, design & selection : PEDS, 25(4), 179-187 (2012-02-16)
Two similar genes, dnmL and rmbA in Streptomyces peucetius, which encode for glucose-1-phosphate (G-1-P) thymidylyltransferases were expressed in Escherichia coli under similar conditions. While RmbA was expressed in soluble form, DnmL was found as insoluble aggregates in inclusion bodies. The
Kyra-Melinda Alexacou et al.
Bioorganic & medicinal chemistry, 18(22), 7911-7922 (2010-10-16)
Glycogen phosphorylase (GP) is a promising target for the treatment of type 2 diabetes. In the process of structure based drug design for GP, a group of 15 aromatic aldehyde 4-(β-d-glucopyranosyl)thiosemicarbazones have been synthesized and evaluated as inhibitors of rabbit
Jef Van der Borght et al.
Biotechnology journal, 5(9), 986-993 (2010-08-28)
β-D-Glucose-1-phosphate (βGlc1P) is an efficient glucosyl donor for both enzymatic and chemical glycosylation reactions but is currently very costly and not available in large amounts. This article provides an efficient production method of βGlc1P from trehalose and phosphate using the
Daniel Decker et al.
Phytochemistry, 79, 39-45 (2012-05-04)
UDP-Glc pyrophosphorylase (UGPase) is an essential enzyme responsible for production of UDP-Glc, which is used in hundreds of glycosylation reactions involving addition of Glc to a variety of compounds. In this study, barley UGPase was characterized with respect to effects
K Stan-Glasek et al.
Folia microbiologica, 55(4), 383-385 (2010-08-04)
Rumen bacterium Pseudobutyrivibrio ruminis strain k3 utilized over 90% sucrose added to the growth medium as a sole carbon source. Zymographic studies of the bacterial cell extract revealed the presence of a single enzyme involved in sucrose digestion. Thin layer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service