Skip to Content
Merck
All Photos(5)

Documents

G7879

Sigma-Aldrich

D-Glucose 6-phosphate sodium salt

≥98% (HPLC)

Synonym(s):

D(+)-Glucopyranose 6-phosphate sodium salt, G-6-P Na, Robison ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12NaO9P
CAS Number:
Molecular Weight:
282.12
Beilstein:
5787568
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥98% (HPLC)

form

crystalline

technique(s)

HPLC: suitable

color

white

useful pH range

4.0-5.0

mp

204 °C (dec.) (lit.)

solubility

water: 50 mg/mL, clear, colorless to very faintly yellow

cation traces

Na: 6.1-10.2% (anhydrous)

storage temp.

room temp

SMILES string

[Na+].O[C@@H]1O[C@H](COP(O)([O-])=O)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C6H13O9P.Na/c7-3-2(1-14-16(11,12)13)15-6(10)5(9)4(3)8;/h2-10H,1H2,(H2,11,12,13);/q;+1/p-1/t2-,3-,4+,5-,6-;/m1./s1

InChI key

ZALKNDISPIVVKC-WYRLRVFGSA-M

Looking for similar products? Visit Product Comparison Guide

Application

D-glucose-6-phosphate sodium can be used as an energy source for Mycobacterium tuberculosis.

Biochem/physiol Actions

In cells, D-glucose 6-phosphate (G6P) is generated when glucose is phosphorylated by hexokinase or glucokinase or by the conversion of glucose-1-phosphate by phosphoglucomutase during glycogenolysis. G6P lies at the beginning of both glycolysis and the pentose phosphate pathways. It also can be stored as glycogen when blood glucose levels are high.

Caution

Non-hygroscopic, and very stable to atmospheric moisture at room temperature, unlike the disodium salt.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iain J Abbott et al.
Antimicrobial agents and chemotherapy, 64(6) (2020-04-08)
There are limited treatment options for enterococcal urinary tract infections, especially vancomycin-resistant Enterococcus (VRE). Oral fosfomycin is a potential option, although limited data are available guiding dosing and susceptibility. We undertook pharmacodynamic profiling of fosfomycin against E. faecalis and E.
Iain J Abbott et al.
The Journal of antimicrobial chemotherapy, 75(4), 988-996 (2019-12-25)
To assess the antibacterial effects of a single 3 g oral fosfomycin dose on Escherichia coli and Klebsiella pneumoniae clinical isolates within a dynamic bladder infection model. An in vitro model simulating dynamic urinary fosfomycin concentrations was used. Target fosfomycin exposure
Iain J Abbott et al.
Journal of microbiological methods, 171, 105861-105861 (2020-02-09)
The impact of the bladder environment on fosfomycin activity and treatment response is uncertain. Standard laboratory media does not reflect the biomatrix of urine, where limited nutritional factors are important for growth and antimicrobial kill rates. We compared fosfomycin activity
Iain J Abbott et al.
Antimicrobial agents and chemotherapy, 64(3) (2020-01-08)
Oral fosfomycin trometamol is licensed as a single oral dose for the treatment of uncomplicated urinary tract infections, with activity against multidrug-resistant uropathogens. The impact of interindividual variability in urinary concentrations on antimicrobial efficacy, and any benefit of giving multiple
Calvin S Carter et al.
Cell metabolism, 32(4), 561-574 (2020-10-08)
Aberrant redox signaling underlies the pathophysiology of many chronic metabolic diseases, including type 2 diabetes (T2D). Methodologies aimed at rebalancing systemic redox homeostasis have had limited success. A noninvasive, sustained approach would enable the long-term control of redox signaling for

Articles

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Protocols

To measure glucose-6-phosphatase activity, the Taussky-Shorr method is used. This method is a spectrophotometric stop-rate determination assay that is measured at 660 nm.

To measure glucose-6-phosphatase activity, the Taussky-Shorr method is used. This method is a spectrophotometric stop-rate determination assay that is measured at 660 nm.

To measure glucose-6-phosphatase activity, the Taussky-Shorr method is used. This method is a spectrophotometric stop-rate determination assay that is measured at 660 nm.

To measure glucose-6-phosphatase activity, the Taussky-Shorr method is used. This method is a spectrophotometric stop-rate determination assay that is measured at 660 nm.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service