Skip to Content
Merck
All Photos(6)

Documents

G4501

Sigma-Aldrich

L-Glutathione oxidized

≥98%

Synonym(s):

GSSG, Glutathiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32N6O12S2
CAS Number:
Molecular Weight:
612.63
Beilstein:
1718700
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

L-Glutathione oxidized, lyophilized powder

application(s)

detection

SMILES string

N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O

InChI

1S/C20H32N6O12S2/c21-9(19(35)36)1-3-13(27)25-11(17(33)23-5-15(29)30)7-39-40-8-12(18(34)24-6-16(31)32)26-14(28)4-2-10(22)20(37)38/h9-12H,1-8,21-22H2,(H,23,33)(H,24,34)(H,25,27)(H,26,28)(H,29,30)(H,31,32)(H,35,36)(H,37,38)/t9-,10-,11-,12-/m0/s1

InChI key

YPZRWBKMTBYPTK-BJDJZHNGSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glu-Cys-Gly, Glu-Cys-Gly [Disulfide bridge: 2a-2b]

Biochem/physiol Actions

L-Glutathione, oxidized is frequently measure in vivo as an indicator of oxidative stress. Oxidized L-Glutathione may be converted to L-glutathione by various reducing systems.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S K Tandon et al.
Toxicology letters, 145(3), 211-217 (2003-10-29)
The influence of an antioxidant agent such as N-acetyl cysteine (NAC) or mannitol on the cadmium chelating ability of monoisoamyl 2,3-dimercaptosuccinate (MiADMS) was investigated in cadmium pre-exposed rats. This ester of 2,3-dimercaptosuccinic acid (DMSA), an accepted drug for lead poisoning
N O Kaakoush et al.
Antimicrobial agents and chemotherapy, 53(5), 1884-1891 (2009-02-19)
Metronidazole resistance in Helicobacter pylori has been attributed to mutations in rdxA or frxA. Insufficient data correlating RdxA and/or FrxA with the resistant phenotype, and the emergence of resistant strains with no mutations in either rdxA or frxA, indicated that
T Kondo et al.
American journal of hematology, 26(1), 37-45 (1987-09-01)
The oxidized form of glutathione transport was studied in human erythrocytes in pyrimidine 5'-nucleotidase (P5N) deficiency, a disorder in which the amounts of CTP and UTP in the erythrocytes are elevated. The inhibition of ATP-requiring oxidized glutathione (GSSG) transport by
Roman Kireev et al.
European journal of pharmacology, 701(1-3), 185-193 (2012-12-12)
Fatty livers occur in up to 20% of potential liver donors and increase cellular injury during the ischemia/reperfusion phase, so any intervention that could enable a better outcome of grafts for liver transplantation would be very useful. The effect of
Janaina Fernandes et al.
Journal of medicinal chemistry, 52(4), 1214-1218 (2009-02-06)
Multidrug resistance-associated protein 1 (MRP1/ABCC1) is a very promiscuous transporter. Herein we used topological polar surface area (TPSA), a descriptor defined as the sum of surfaces of polar atoms in a molecule, to analyze drug transport by MRP1. We suggested

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service