Skip to Content
Merck
All Photos(1)

Key Documents

G4507

Sigma-Aldrich

Giemsa stain

Technical grade, powder

Synonym(s):

Azure mixture sicc. Giemsa stain

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14ClN3S
CAS Number:
Molecular Weight:
291.80
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Giemsa stain, technical grade, used as a blood stain

grade

technical grade

form

powder

color

dark greenvery dark

mp

300 °C (lit.)

solubility

ethanol: 1 mg/mL

λmax

645-655 nm

suitability

application for blood stain

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].CN(C)c1ccc2N=C3C=CC(=[NH2+])C=C3Sc2c1

InChI

1S/C14H13N3S.ClH/c1-17(2)10-4-6-12-14(8-10)18-13-7-9(15)3-5-11(13)16-12;/h3-8,15H,1-2H3;1H

InChI key

NALREUIWICQLPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Giemsa stain is a versatile differential stain that is widely used in hematology for blood and bone marrow specimens, clinical cytology specimens, bacteriology, histological biopsies, and tumor samples. Giemsa’s staining solution is composed of methylene blue, azure, and eosin. It is a major histological stain due to its unique staining of chromatin, nuclear membranes, and cytoplasmic elements. The stain is frequently used in combination with other dye solutions: May-Grünwald’s solution for Pappenheim (MGG) and Wright-Giemsa.

Application

Giemsa stain has been used for the staining of cells and blood films.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amira Al-Hosary et al.
Parasites & vectors, 13(1), 495-495 (2020-10-01)
Anaplasma marginale is an obligate intracellular bacterium and the main cause of bovine anaplasmosis in tropical and subtropical regions. In Egypt, data regarding the prevalence of A. marginale in ruminant hosts and of the circulating genotypes is lacking. This study
Hao Li et al.
OncoTargets and therapy, 13, 1211-1223 (2020-02-28)
6-thioguanine (6-TG), as a conventional "ancient" drug for the treatment of acute leukemia, has been proved to have extensive anti-tumor roles. This study was created to investigate the hidden function of 6-TG on the MCF-7 breast cancer cell line (ER+
Kevin M Swift et al.
Sleep, 43(5) (2019-12-01)
Sleep impacts diverse physiological and neural processes and is itself affected by the menstrual cycle; however, few studies have examined the effects of the estrous cycle on sleep in rodents. Studies of disease mechanisms in females therefore lack critical information
Xianhua Yin et al.
Applied and environmental microbiology, 75(18), 5779-5786 (2009-07-28)
O island 48 (OI-48) of Escherichia coli consists of three functional gene clusters that encode urease, tellurite resistance (Te(r)), and putative adhesins Iha and AIDA-1. The functions of these clusters in enterohemorrhagic E. coli (EHEC) O157:H7 infection are unknown. Deletion
Iset Medina Vera et al.
PLoS pathogens, 7(9), e1002197-e1002197 (2011-09-13)
Apicomplexans are obligate intracellular parasites that invade host cells by an active process leading to the formation of a non-fusogenic parasitophorous vacuole (PV) where the parasite replicates within the host cell. The rhomboid family of proteases cleaves substrates within their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service