Skip to Content
Merck
All Photos(3)

Key Documents

G5637

Sigma-Aldrich

Giemsa stain

certified by the Biological Stain Commission, powder or crystals

Synonym(s):

Azure mixture sicc. Giemsa stain

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14ClN3S
CAS Number:
Molecular Weight:
291.80
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

Giemsa stain, certified by the Biological Stain Commission

grade

certified by the Biological Stain Commission

Quality Level

form

powder or crystals

technique(s)

microbe id | staining: suitable

color

dark green to very dark green

mp

300 °C (lit.)

solubility

ethanol: 1 mg/mL, blue to very deep blue

absorption

≥0.6 at 647-653 nm in methanol at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].CN(C)c1ccc2N=C3C=CC(=[NH2+])C=C3Sc2c1

InChI

1S/C14H13N3S.ClH/c1-17(2)10-4-6-12-14(8-10)18-13-7-9(15)3-5-11(13)16-12;/h3-8,15H,1-2H3;1H

InChI key

NALREUIWICQLPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Giemsa is a versatile polychromatic stain, which is suitable for staining a diverse range of specimens. It is a buffered thiazine-eosinate solution designed to provide coloration of blood cells. Giemsa’s stain is frequently used for diagnostic purposes in hematology to differentiate the nuclear and cytoplasmic morphology of platelets, RBCs, WBCs, and parasites. It is frequently used in combination with other dye solutions as May-Grunwald’s solution for Pappenheim (MGG) and Wright-Giemsa. The Giemsa stain was adapted to histology due to its unique staining of chromatin, nuclear membranes, and cytoplasmic elements. Giemsa′s staining solution is composed of methylene blue, azure, and eosin.

Application

Giemsa stain has been used for the staining of cells and blood films.[1][2]It is required for the staining of azurophil granules and is an excellent stain for inclusion bodies.[3]Its applications can be seen in hematology for blood and bone marrow specimens, bacteriology, clinical cytology specimens, histological biopsies, and tumor samples.

Suitability

Certified for Giemsa stain for blood films and for the Wolbach′s Giemsa method for paraffin sections.

Other Notes

Nuclei will be varying shades of purple. Cytoplasmic staining will be varying shades of blue to light pink. Fine reddish to lilac granules may be present in cytoplasm of some cell types. Basophils will demonstrate dark blue black granules in the cytoplasm. Eosinophils will demonstrate bright orange granules in the cytoplasm. Red blood cells should be pink to orange

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The interaction between Plasmodium falciparum and P. vivax in children on Espiritu Santo island, Vanuatu.
Maitland K et al.
Transactions of the Royal Society of Tropical Medicine and Hygiene, 90, 614-614 (1996)
Hui Tong et al.
Cancer management and research, 11, 1869-1879 (2019-03-19)
Hepatocellular carcinoma (HCC) is the third leading cause of death by malignancy worldwide. HCC has a poor prognosis due to tumor invasiveness and metastasis. There is substantial evidence that the epithelial-to-mesenchymal transition (EMT) plays a central role in cancer metastasis.
Mollie Jurewicz et al.
Diabetes, 59(12), 3139-3147 (2010-09-16)
A number of clinical trials are underway to test whether mesenchymal stem cells (MSCs) are effective in treating various diseases, including type 1 diabetes. Although this cell therapy holds great promise, the optimal source of MSCs has yet to be
Xianhua Yin et al.
Applied and environmental microbiology, 75(18), 5779-5786 (2009-07-28)
O island 48 (OI-48) of Escherichia coli consists of three functional gene clusters that encode urease, tellurite resistance (Te(r)), and putative adhesins Iha and AIDA-1. The functions of these clusters in enterohemorrhagic E. coli (EHEC) O157:H7 infection are unknown. Deletion
Iset Medina Vera et al.
PLoS pathogens, 7(9), e1002197-e1002197 (2011-09-13)
Apicomplexans are obligate intracellular parasites that invade host cells by an active process leading to the formation of a non-fusogenic parasitophorous vacuole (PV) where the parasite replicates within the host cell. The rhomboid family of proteases cleaves substrates within their

Questions

  1. Hello, could you confirm the dye(s) present in G5367? Is it only the pictured compound (I think Azure A) without methylene blue or eosin? Thank you!

    1 answer
    1. Giemsa stain, as defined by the Biological Stain Commission, consists of a polychrome methylene blue and eosin y mixture. When methylene blue undergoes the polychroming process, various homologs of methylene blue are expected to be present. Azure A, Azure B, Azure C, and other homologs may be present. The percentages of the various homologs can and will vary. This product is not tested to determine the identity or concentration of these homologs.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service