Skip to Content
Merck
All Photos(2)

Key Documents

MG500

Sigma-Aldrich

May-Grünwald Stain

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116121
NACRES:
NA.47

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

0.25 % (w/v) in methanol

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

May-Grünwald Stain has been employed to stain neutrophil granules and granulocyte inclusion bodies. May-Grünwald-Giemsa might be useful in the diagnosis of Malassezia folliculitis, a fungal mediated disorder.

Application

For use in the differential staining of cellular elements of blood

Preparation Note

Prepared with non-polychromed (non-oxidized) methylene blue and eosin Y.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuria Castro-Bravo et al.
Frontiers in microbiology, 8, 1405-1405 (2017-08-10)
An extracellular layer of exopolysaccharides (EPS) covers the surface of some
Resveratrol induces human K562 cell apoptosis, erythroid differentiation, and autophagy.
Yan HW, Hu WX, Zhang JY, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 35(6), 5381-5388 (2014)
Jianbing Zhang et al.
PLoS genetics, 14(9), e1007665-e1007665 (2018-09-25)
Heme-iron recycling from senescent red blood cells (erythrophagocytosis) accounts for the majority of total body iron in humans. Studies in cultured cells have ascribed a role for HRG1/SLC48A1 in heme-iron transport but the in vivo function of this heme transporter
Tomasz Dobrzycki et al.
Communications biology, 3(1), 71-71 (2020-02-15)
Gata2 is a key transcription factor required to generate Haematopoietic Stem and Progenitor Cells (HSPCs) from haemogenic endothelium (HE); misexpression of Gata2 leads to haematopoietic disorders. Here we deleted a conserved enhancer (i4 enhancer) driving pan-endothelial expression of the zebrafish
Epidemiological characteristics of Malassezia folliculitis and use of the May-Grunwald-Giemsa stain to diagnose the infection
Durdu, et al.
Diagnostic Microbiology and Infectious Disease, 76(4) (2013)

Related Content

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service