Skip to Content
Merck
All Photos(1)

Key Documents

G3000

Sigma-Aldrich

β-Gentiobiose

≥85% (remainder primarily α-anomer)

Synonym(s):

6-O-β-D-Glucopyranosyl-D-glucose, Amygdalose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
93354
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Quality Level

Assay

≥85% (remainder primarily α-anomer)

form

powder

optical activity

[α]20/D 8.5 to 10.5, c = 2% (w/v) in water

color

beige

solubility

water: 100 mg/mL, clear to slightly hazy, colorless to light yellow (

)

storage temp.

room temp

SMILES string

OC[C@H]1O[C@@H](OC[C@H]2OC(O)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-3-5(14)8(17)10(19)12(23-3)21-2-4-6(15)7(16)9(18)11(20)22-4/h3-20H,1-2H2/t3-,4-,5-,6-,7+,8+,9-,10-,11?,12-/m1/s1

InChI key

DLRVVLDZNNYCBX-CQUJWQHSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Gentiobiose was used in binding aβays to determine if coelomic cytolytic factor 1 (CCF-1) binds to β-1,3-glucans to stimulate the pro-PO-activating system.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cosima Damiana Calvano et al.
Journal of the American Society for Mass Spectrometry, 28(8), 1666-1675 (2017-05-04)
The superbasic proton sponge 1,8-bis(tripyrrolidinylphosphazenyl)naphthalene (TPPN) has been successfully employed for the structural characterization of neutral saccharides, cyclodextrins, and saccharide alditols by matrix assisted laser desorption/ionization tandem mass spectrometry (MALDI-MS/MS). Owing to its inherently high basicity, TPPN is capable of
Rikuto Ishiguro et al.
FEBS letters, 591(23), 3926-3936 (2017-11-14)
BT_3567 protein, a putative β-glucosidase from Bacteroides thetaiotaomicron, exhibits higher activity toward Sop
Ayla Sant'Ana da Silva et al.
Biotechnology letters, 39(11), 1717-1723 (2017-08-03)
Glucose conversion into disaccharides was performed with β-glucosidases from Prunus dulcis (β-Pd), Aspergillus niger (β-An) and A. awamori (β-Aa), in reactions containing initial glucose of 700 and 900 g l The reactions' time courses were followed regarding glucose and product concentrations. In
A Beschin et al.
The Journal of biological chemistry, 273(38), 24948-24954 (1998-09-12)
Coelomic fluid of Eisenia foetida earthworms contains a 42-kDa protein named coelomic cytolytic factor 1 (CCF-1) that was described previously to be involved in cytolytic, opsonizing, and hemolytic properties of the coelomic fluid. Cloning and sequencing of CCF-1 reveal significant
Arun Nair et al.
PloS one, 8(4), e60538-e60538 (2013-04-18)
The triglucoside of sesaminol, i.e., 2,6-O-di(β-D-glucopyranosyl)-β-D- glucopyranosylsesaminol (STG), occurs abundantly in sesame seeds and sesame oil cake and serves as an inexpensive source for the industrial production of sesaminol, an anti-oxidant that displays a number of bioactivities beneficial to human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service